Skip to Content
Merck
All Photos(1)

Key Documents

03732738001

Roche

Dideoxynucleoside Triphosphate Set

Sequencing Grade, sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21

grade

Molecular Biology

Quality Level

Assay

≤1.5% (ddNDP, HPLC)
≥98% (ddNTP, HPLC)

form

liquid

packaging

pkg of 4 × 100 μL (4 x 1 μmol; 10 mM, each)

manufacturer/tradename

Roche

color

colorless

solubility

water: miscible

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

The Dideoxynucleoside Triphosphate Set, Sequencing Grade, sodium salt solution, consists of dideoxynucleotides of very high purity (ddNTP HPLC, area% ≥98%; ddNDP HPLC, area% ≤1.5%), specifically manufactured and tested for sequencing reactions.
The set contains 4 individual vials of ddATP, ddCTP, ddGTP, and ddTTP, each at 10 mM as a clear, colorless solution of the sodium salt (pH 8.3).

Application

2′,3′-Dideoxynucleoside triphosphates inhibit the chain elongation of a given primer catalyzed by the DNA polymerase (e.g., Klenow enzyme) and are therefore used for DNA sequencing according to Sanger (Sanger, F. et al. (1977) PNAS USA 74, 5463).
Sequencing is achieved by including in each reaction a dideoxynucleotide that acts as a chain terminator. Four reactions are set up, each containing the same template and primer, and a chain terminator specific for A, C, G, or T. Because only a small amount of the chain terminator is included, incorporation into the new DNA strand is a random event. Each reaction therefore generates a collection of fragments, and every DNA strand will end at the same type of base (A, C, G, or T).

Packaging

The set contains 4 individual vials of ddATP, ddCTP, ddGTP and ddTTP each at 10 mM as a clear, colorless solution of the sodium salt (pH 8.3).

Quality

Each lot of dideoxynucleotide solution is assayed for function in sequencing reaction.
Tested for absence of contaminating deoxyribonucleases, ribonucleases and nicking activities according to the current Quality Control procedures.

Preparation Note

Activator: Mg2+, Co2+ and Mn2+ ions
Working solution: Preparation of Termination Mix
Before preparing the termination mix, dilute an appropriate volume of each dideoxynucleotide in water to a concentration of 10 μM.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • ddATP, sodium salt 10 mM

  • ddCTP, sodium salt 10 mM

  • ddGTP, sodium salt 10 mM

  • ddTTP, sodium salt 10 mM

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted elimination of mutated mitochondrial DNA by a multi-functional conjugate capable of sequence-specific adenine alkylation
Hidaka, et al.
Cell Chemical Biology, 29, 690-695 (2021)
Atypical UV Photoproducts Induce Non-canonical Mutation Classes Associated with Driver Mutations in Melanoma
Laughery, et al.
Cell Reports, 33 (2020)
Staphylococcus aureus 30S Ribosomal Subunit Purification and Its Biochemical and Cryo-EM Analysis
Belinite, et al.
Bio-protocol, 12 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service