ZFC0NN5QQ
Connector 5m
Connects two PODs, for Milli-Q® IQ/IX series
Synonym(s):
Connector 5m POD to POD
About This Item
feature
For POD to POD connection
cable L
5 m
compatibility
for use with E-POD®
for use with Milli-Q® IQ Element
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
for use with Q-POD®
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service