Skip to Content
Merck
All Photos(1)

Documents

XXKT0471J

Millipore

VACUUM FILT KIT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service