XX2504701
Filter holder support for manifold, stainless steel
Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.24
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
input
sample type beer
sample type pharmaceutical(s)
sample type bottled water
sample type cosmetics
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
Related Categories
General description
stainless steel manifolds for sterility testing.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service