Yes, the Millex High Output and Chemical Duty pumps are CE approved.
WP6122050
Chemical Duty Pump, 220 V/50 Hz
Synonym(s):
Laboratory vacuum pump
About This Item
Recommended Products
AC/DC input
220 V, 50 Hz
shipped in
ambient
Application
Features and Benefits
- High flow rates enabling minimal filtration time, Portable design enables sharing between workstations.
- Vacuum source for filter holders, manifolds, and devices.
- Used with Samplicity® Filtration System, SNAP id® 2.0 System, Multiscreen® HTS Filtration Manifold, and Stericup® Filters.
Legal Information
replacement part
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is product WP6122050 CE Approved?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service