WBLUC
Immobilon® Classico Western HRP substrate
Western blotting, dot/slot blotting
Synonym(s):
HRP substrate, chemiluminescent detection substrate, detection substrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22
Recommended Products
Product Name
Immobilon Classico Western HRP substrate, Western blotting, dot/slot blotting
conjugate
peroxidase conjugate
Quality Level
manufacturer/tradename
Luminata
technique(s)
western blot: suitable
membrane area
1000 cm2 , membrane coverage
detection method
chemiluminescent
shipped in
ambient
General description
Premixed, ready to use chemiluminescent HRP detection reagent
Application
For use on immunoblots.
Western blotting, dot/slot blotting
Packaging
1 bottle for 1000 cm2
Quality
The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.
Storage and Stability
Stable for 1 year at 4°C from date of shipment. Contains hydrogen peroxide. Use in well ventilated area.
Legal Information
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Implication of RPA32 phosphorylation in S-phase checkpoint signalling at replication forks stalled with aphidicolin in Xenopus egg extracts.
Recolin, Benedicte and Maiorano, Domenico
Biochemical and biophysical research communications (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service