Skip to Content
Merck

VPMD101NB1

Millipore

Viresolve® Pro Device

Parvovirus removal filter, Modus 1.1 format, filtration area 0.017 m²

Synonym(s):

Viresolve® Pro Device Modus 1.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

material

PVDF housing
polyethersulfone membrane
silicone gasket

Quality Level

product line

EMPROVE® Filter

manufacturer/tradename

Viresolve®

parameter

60 psig max. inlet pressure

dilution

(Scale studies/Pilot)

H

5.92 cm (2.33 in.)

L

18.62 cm (7.33 in.)

W

9.22 cm (3.63 in.)

device size

1.1 in.

filtration area

0.017 m2

impurities

<0.25 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

fitting

1/4 in. integrated vent hose barb (with double O-ring seal)
3/4 in. sanitary TC inlet/outlet fitting(s)

Looking for similar products? Visit Product Comparison Guide

General description

The Viresolve® Pro Solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.

Viresolve Pro Modus devices are typically used for filtration of pilot to mid-scale batch volumes of proteins. The Viresolve® Pro Device Modus 1.1 offers a filtration area of 0.017 m2 and has a primary use in pilot/small-volume processing.

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.
Directions for Use

  • Organism Retention: Parvovirus
  • Mode of Action: Filtration (size exclusion)
  • Application: Protein purification
  • Intended Use: Viral clearance
  • Instructions for Use: Please see the user guide shipped with this product
  • Storage Statement: Store at room temperature
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Preparation Note

This product was manufactured with materials that meet the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Analysis Note

TOC/Conductivity
After a controlled water flush of 50 L/m², samples exhibited less than 500 ppb TOC per USP <64> and less than 1.3 µS/cm per USP <64>.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

EMD Millipore Corporation certifies that this product complies with the European Pressure Equipment Directive, 97/23/EC of 29 May 1997. This product has been classified under Article 3 § 3 of the Pressure Vessel Directive. It has been designed and manufactured in accordance with sound engineering practice to ensure safe use. In compliance with Article 3 § 3 of this Pressure Vessel Directive, this product does not bear the CE mark.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service