UFC701008
Centricon® Plus Centrifugal Filter
MWCO 10 kDa, sample volume 15-70 mL, regenerated cellulose membrane
Synonym(s):
Centrifuge concentrator
About This Item
Recommended Products
material
polypropylene cap
polypropylene cup (for concentrate/retentate)
polypropylene cup (for filtrate collection)
polypropylene cup (for sample filter)
regenerated cellulose membrane
Quality Level
sterility
non-sterile
product line
Amicon®
feature
holdup volume 75 μL
manufacturer/tradename
Amicon® Ultra
Centricon®
parameter
15-70 mL sample volume
technique(s)
protein extraction: suitable
protein purification: suitable
L
121 mm
diam.
6 cm
filtration area
19 cm2
pore size
10 kDa MWCO
application(s)
sample preparation
shipped in
ambient
General description
Application
- Concentrating and desalting chromatography column eluates
- Concentrating monoclonal antibodies
- Concentrating proteins or viruses from culture supernatants
- Clarifying tissue homogenates and cell lysates
- Buffer exchange or diafiltration
Features and Benefits
- 90% typical recovery
- 50X to 200X concentration
- Low hold-up volume
- Polypropylene housing minimizes binding
- Can concentrate most 70 mL solutions down to 350 μL in just 25 minutes
- Dead stop prevents spinning to dryness
- Ready to use. No assembling is required.
Physical form
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service