UFC30GV0S
Ultrafree® Centrifugal Filter, 0.5 mL Sample Volume
pore size 0.22 μm, PVDF membrane (hydrophilic)
Synonym(s):
Centrifuge filter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24
Recommended Products
material
PVDF membrane (hydrophilic)
polypropylene
Quality Level
sterility
sterile
feature
holdup volume 5 μL
manufacturer/tradename
Ultrafree®
parameter
0.5 mL sample volume
50 °C max. temp.
technique(s)
protein extraction: suitable
protein purification: suitable
L
45 mm
diam.
10.6 mm
filtration area
0.2 cm2
matrix
Durapore®
pore size
0.22 μm pore size
application(s)
sample preparation
shipped in
ambient
Related Categories
General description
Ultrafree®-MC (0.5 mL) and Ultrafree®-CL (2 mL) centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes (from 0.1 to 5.0 m) for fast filtration and highly reproducible performance. Pre-sterilized units are also available. Use in fixed-angle rotors for 1.5 mL tubes (-MC) and 15 mL tubes (CL).
Filter Code: GVPP
Filter Type: Microfiltration
Filter Code: GVPP
Filter Type: Microfiltration
Ultrafree®-MC (0.5 mL) and Ultrafree®-CL (2 mL) centrifugal devices are single-use, disposable filters used for removing particulates from aqueous biological solutions. These devices are available in two processing volumes with a range of microporous membranes (from 0.1 to 5.0 m) for fast filtration and highly reproducible performance. Pre-sterilized units are also available. Use in fixed-angle rotors for 1.5 mL tubes (-MC) and 15 mL tubes (CL).
Filter Code: GVPP
Filter Type: Microfiltration
Filter Code: GVPP
Filter Type: Microfiltration
Application
Ultrafree®-MC GV Centrifugal Filter has been used
- to filter cell lysates after centrifugation for Label-free mass spectrometry
- for filtering extracted enzymes in the buffer for preparing imaging solution for cellular direct stochastic optical reconstruction microscopy (dSTORM) and DNA-based point accumulation for imaging in nanoscale topography (DNA-PAINT) imaging
- to purify viral solution during transfection studies
- to filter heart tissue homogenates for preparation of the sample for high-pressure liquid chromatography (HPLC) analysis
Features and Benefits
- Used for removal of particles and precipitates from aqueous and some solvent-based samples
- Ideal for use with protein and nucleic acid solutions
- Used for removal of particles and precipitates from aqueous and some solvent-based samples
- Ideal for use with protein and nucleic acid solutions
Physical form
Color Code: Yellow
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Ultrafree is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Beta2-glycoprotein I can exist in 2 conformations: implications for our understanding of the antiphospholipid syndrome.
Agar, Cetin, et al.
Blood, 116, 1336-1343 (2010)
Robert Player et al.
BMC genomics, 21(1), 166-166 (2020-02-19)
The state-of-the-art in nucleic acid based biodetection continues to be polymerase chain reaction (PCR), and many real-time PCR assays targeting biodefense pathogens for biosurveillance are in widespread use. These assays are predominantly singleplex; i.e. one assay tests for the presence
Cetin Agar et al.
Blood, 117(25), 6939-6947 (2011-04-02)
Sepsis is a systemic host response to invasive infection by bacteria. Despite treatment with antibiotics, current mortality rates are in the range of 20%-25%, which makes sepsis the most important cause of death in intensive care. Gram-negative bacteria are a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service