SCT153
Renilla Luciferase Assay
Flash-type Renilla Luciferase assay designed to yield reliable, linear measurements of Renilla luciferase activity with minimal autoluminescence background and superior sensitivity.
Synonym(s):
Firefly Luciferase Assay, Bioluminescent Assay
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
cell based assay: suitable
detection method
chemiluminescent
shipped in
ambient
General description
Renilla luciferase has been used as a reporter gene for studying gene regulation and function in vitro and in vivo. It commonly is used in multiplex transcriptional reporter assays or as a normalizing transfection control for firefly luciferase assays. The enzyme does not require post-translational modification for its activity and may function as a genetic reporter immediately following translation. Coelenterazine also emits light from enzyme-independent oxidation, a process known as autoluminescence. The autoluminescence is enhanced by superoxide anion and peroxynitrite in cells and tissues.
The Renilla Luciferase Assay utilizes a special buffer formulation designed to yield reliable, linear measurements of Renilla luciferase activity with minimal autoluminescence background and superior sensitivity. This is a flash-type luminescence assay that requires signal to be measured immediately after adding working solution to samples. The luminescence signal decays over the course of about 2 minutes of reaction time, although signal half-life may vary depending on luciferase expression levels.
The Renilla Luciferase Assay utilizes a special buffer formulation designed to yield reliable, linear measurements of Renilla luciferase activity with minimal autoluminescence background and superior sensitivity. This is a flash-type luminescence assay that requires signal to be measured immediately after adding working solution to samples. The luminescence signal decays over the course of about 2 minutes of reaction time, although signal half-life may vary depending on luciferase expression levels.
Application
Firefly Luciferase Assay
Flash-type Renilla Luciferase assay designed to yield reliable, linear measurements of Renilla luciferase activity with minimal autoluminescence background and superior sensitivity.
Research Category
Cell Signaling
Cell Signaling
Research Sub Category
Live Cell Dye
Live Cell Dye
Components
1) 5X Passive Lysis Buffer (CS224582): 30 mL
2) Renilla Luciferase Assay Buffer 2.0 (CS224583): 100 mL
3) Aquaphile Coelenterazine (CS224584): 1 X 4 mg
2) Renilla Luciferase Assay Buffer 2.0 (CS224583): 100 mL
3) Aquaphile Coelenterazine (CS224584): 1 X 4 mg
Physical form
liquid
Storage and Stability
Store Renilla Luciferase Assay at –80°C. Renilla Luciferase Assay Buffer 2.0 is stable at -80°C for at least six months from date of receipt. Other kit components are stable at -20°C for at least six months from date of receipt. Kit components and stock solutions of Aquaphile coelenterazine in water are stable to at least 5 freeze/thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service