SCT010M
LuminiCell Tracker 540- Cell Labeling Kit
Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to label cells for long term cell tracking and cell tracing in vitro and in vivo experiments.
Synonym(s):
AIEDots Cell Tracker Dye, AIE Live Cell Fluorescent Dye, Fluorescent Cell Tracking Dye, Live Cell Fluorescent Nanoparticles
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
General description
Absorbance Maximum: 423nm
Emisson Maximum: 540nm
Suggested Filters:
Excitation: 405/458/488nm
Emisson: 480-560nm
Emisson Maximum: 540nm
Suggested Filters:
Excitation: 405/458/488nm
Emisson: 480-560nm
Long-term noninvasive cell tracking by fluorescent probes and quantum dots is of great importance to life science and biomedical engineering. Current methods used to fluorescently tag cells have been limited by short signal duration, high background auto-fluorescence or lengthy molecular cloning manipulations using GFP. LuminiCell Trackers<TMSYMBOL></TMSYMBOL> are biocompatible organic fluorescent nanoparticles based on Aggregation Induced Emission technology, termed AIEDots. Aggregation induced emission (AIE) molecules emit fluorescence in an opposite manner than other common fluorophores (Quantum Dots, GFP). Propeller-shaped AIE fluorogens are non-emissive in solutions but become highly fluorescent upon aggregate formation. Due to these differences, LuminiCell Trackers<TMSYMBOL></TMSYMBOL> have very high fluorescence intensities with minimal signal quenching allowing live cell fluorescent tagging for up to 10 days in vitro and 21 days in vivo. These properties make them optimal candidates for long interval live cell bioimaging experiments.
The Luminicell Tracker<TMSYMBOL></TMSYMBOL>-540 Cell Labeling Kit contain green fluorescent AIEDot nanoparticles with TAT sequences. These nanoparticles can be used to fluorescently tag cells for long term cell tracking and tracing experiments.
Features and Benefits
● Brighter: 10X brighter than other cell labeling technologies
● Photostable: 3X longer fluorescence without signal quenching
● Biocompatible: Non-toxic organic nanoparticles intended for biological applications
● Rapid Protocol: Easy-to-use protocol labels cells within 4 hours
The Luminicell Tracker<TMSYMBOL></TMSYMBOL>-540 Cell Labeling Kit contain green fluorescent AIEDot nanoparticles with TAT sequences. These nanoparticles can be used to fluorescently tag cells for long term cell tracking and tracing experiments.
Features and Benefits
● Brighter: 10X brighter than other cell labeling technologies
● Photostable: 3X longer fluorescence without signal quenching
● Biocompatible: Non-toxic organic nanoparticles intended for biological applications
● Rapid Protocol: Easy-to-use protocol labels cells within 4 hours
Application
Biocompatible organic fluorescent nanoparticles (AIEDots) that can be used to label cells for long term cell tracking and cell tracing in vitro and in vivo experiments.
Quality
Absorbance: 422+-5nm
Concentration: 180-220nM
Fluorescence: 540+/- 10nm
Quantum Yield: ≥50%
Brightness at 540nM: ≥3X10^7 M-1cm-1
Cellular Assay: HeLa Cell Fluorescence
Concentration: 180-220nM
Fluorescence: 540+/- 10nm
Quantum Yield: ≥50%
Brightness at 540nM: ≥3X10^7 M-1cm-1
Cellular Assay: HeLa Cell Fluorescence
Physical form
20nM in 0.5ml PBS, pH 7.4
Storage and Stability
Store at -2-8°C upon receipt. Thaw at room temperature or in a water bath. Do not Freeze.
Note: Some particulates may form as a result of nanoparticle aggregation during shipping. To get particulates back in solution, sonicate the vial containing LuminiCell Tracker three times for 1 min each before use.
Note: Some particulates may form as a result of nanoparticle aggregation during shipping. To get particulates back in solution, sonicate the vial containing LuminiCell Tracker three times for 1 min each before use.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Long-Term Tracking Mesenchymal Stem Cell Differentiation with Photostable Fluorescent Nanoparticles.
ACS applied materials & interfaces, 8(19), 11925-11933 (2016-04-29)
Mesenchymal stem cells (MSCs) have proved to be a promising and abundant cell source for tissue and organ repair in regenerative medicine. However, the cell fate, distribution and migration of these transplanted cells are still unclear due to the limited
Aggregation-Induced Emission: Together We Shine, United We Soar!
Chemical reviews, 115(21), 11718-11940 (2015-10-23)
Scientific reports, 3, 1150-1150 (2013-01-30)
Long-term noninvasive cell tracing by fluorescent probes is of great importance to life science and biomedical engineering. For example, understanding genesis, development, invasion and metastasis of cancerous cells and monitoring tissue regeneration after stem cell transplantation require continual tracing of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service