Skip to Content
Merck
All Photos(2)

Key Documents

SCNY00060

Millipore

Millipore® Steriflip® Vacuum Tube Top Filter

pore size 60.0 μm, Nylon membrane (net), funnel capacity 50 mL, pack of 25 ea

Synonym(s):

Steriflip® Nylon Net Sterile Centrifuge Tube Top Filter Unit, tube top sterile filter unit, tube top vacuum filter, tube top vacuum sterile filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031602
NACRES:
NB.22

material

Nylon membrane (net)
acrylic housing
polypropylene

Quality Level

sterility

sterile

product line

Steriflip®

feature

holdup volume 0.6 mL
hydrophilic

packaging

pack of 25 ea

parameter

45 °C max. temp.

technique(s)

sterile filtration: suitable

H

138 mm

capacity

50 mL

diam.

6.4 cm

filter diam.

4 cm

filtration area

7 cm2

funnel capacity

50 mL

pore size

60.0 μm pore size

fitting

double lead thread inlet (with vacuum port)
double lead thread outlet for 50 mL centrifuge tube

shipped in

ambient

General description

The Steriflip® filter unit is designed to work with standard 50 mL centrifuge tubes, which eliminates sample transfer. It is useful for sterilization of aqueous solutions (up to 50 ml), sterile filtration of small batches of culture media, sterile filtration of buffers with high-value protein or small molecule components, and cell isolation.

Application

For Research Use Only

Steriflip® Sterile Centrifuge Tube Top Filter Unit has been used in the isolation of umbilical cord mesenchymal stromal cells and fibroblast and smooth muscle cells.

Features and Benefits

  • Eliminates messy sample transfer via a simple vacuum-assisted, closed system
  • Can isolate cells from cellular debris using any of the various pore-sized nylon net filters
  • Ideal for sterilizing small batches of culture media
  • Attaches to any standard 50 mL centrifuge tube, eliminating unnecessary materials, time, and risk of spillage

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Steriflip is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Gao et al.
Frontiers in cell and developmental biology, 9, 630228-630228 (2021-04-17)
Non-invasive biomarkers to identify patients with bladder outlet obstruction (BOO)-related dysfunction are still needed to guide clinical practice. The current study aims to investigate molecular alterations and biomarkers associated with partial BOO (PBOO) in rats. Sprague-Dawley rats were used to
J Robert Smith et al.
Stem cells international, 2016, 6810980-6810980 (2016-03-12)
Umbilical cord derived mesenchymal stromal cells (UC-MSCs) are a focus for clinical translation but standardized methods for isolation and expansion are lacking. Previously we published isolation and expansion methods for UC-MSCs which presented challenges when considering good manufacturing practices (GMP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service