Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
monoclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
11
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
clone
monoclonal
manufacturer/tradename
Chemicon®
concentration
2.2 mg/mL
technique(s)
ELISA: suitable
shipped in
dry ice
target post-translational modification
unmodified
General description
Purity: 95% or greater
Application
Mouse IgG2a Monoclonal Antibody control validated for use in ELISA.
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
Physical form
Format: Purified
Purified immunoglobulin. Liquid containing no preservatives.
Preparation Note
Maintain at -20°C in undiluted aliquots for up to 12 months. Do not thaw and refreeze.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The vitamin E analog, alpha-tocopheryloxyacetic acid enhances the anti-tumor activity of trastuzumab against HER2/neu-expressing breast cancer.
Hahn, T; Bradley-Dunlop, DJ; Hurley, LH; Von-Hoff, D; Gately, S; Mary, DL; Lu et al.
BMC Cancer null
Martin Bentler et al.
Molecular therapy. Methods & clinical development, 30, 576-592 (2023-09-11)
De novo immune responses are considered major challenges in gene therapy. With the aim to lower innate immune responses directly in cells targeted by adeno-associated virus (AAV) vectors, we equipped the vector capsid with a peptide known to interfere with
A conserved anti-DNA antibody idiotype associated with nephritis in murine and human systemic lupus erythematosus.
R H Weisbart, D T Noritake, A L Wong, G Chan, A Kacena, K K Colburn, R H Weisbart et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Aberrant activation of the interleukin-2 autocrine loop through the nuclear factor of activated T cells by nonleukemogenic human T-cell leukemia virus type 2 but not by leukemogenic type 1 virus.
Niinuma, A; Higuchi, M; Takahashi, M; Oie, M; Tanaka, Y; Gejyo, F; Tanaka, N; Sugamura et al.
Journal of virology null
Richard H Weisbart et al.
Molecular immunology, 39(13), 783-789 (2003-03-06)
Methods for cell type specific targeted intracellular delivery of proteins in vivo remain limited. A murine monoclonal anti-dsDNA antibody, mAb 3E10, was selectively transported into skeletal muscle cells in vivo. The antibody bound a 200 kDa protein only found in
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PP102 | 04053252502972 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service