Skip to Content
Merck
All Photos(3)

Key Documents

PIHP03050

Millipore

Millicell® Standing Cell Culture Inserts

pore size 0.4 μm, diam. 30 mm, translucent polycarbonate membrane, size 6 wells, Tissue Culture (TC)-treated surface, sterile

Synonym(s):

Millicell Cell Culture Insert, 30 mm, polycarbonate, 0.4 µm, Millicell-PCF, cell culture inserts, permeable culture insert, plate inserts, tissue culture insert, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polystyrene housing
translucent polycarbonate membrane

Quality Level

sterility

γ-irradiated
sterile

feature

hydrophilic
100x106 pores/cm2 (pore density)

packaging

pack of 50

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable
drug transporter assay: suitable
permeability assay: suitable

H

13 mm

diam.

30 mm

size

6 wells

surface area

4.2 cm2

working volume

1.5 mL

color

translucent membrane

matrix

Isopore

pore size

0.4 μm

binding type

Tissue Culture (TC)-treated surface

shipped in

ambient

General description

Device Configuration: 6-well plate, 0.4μm pore size

With Millicell® inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell® inserts make it possible to study both sides of the cell monolayer.

Millicell® Standing Inserts:
- Promotes excellent cell growth and provides an exceptional opportunity for cell studies
- Available with Biopore (PTFE) membrane, MF-Millipore (mixed cellulose esters) membrane, and polycarbonate membrane

Membrane Type:
Isopore Membrane (polycarbonate)
- For growth of attachment-dependent cells without matrix

Applications
Cell Attachment, Cell Growth, Cell Differentiation , Immunocytochemistry

Application

Polycarbonate cell culture insert with pore size of 0.4 µm used in a 6-well plate for cell attachment, cell culture, cell differentiation, drug transport & permeability.
Research Category
Cell Culture

Packaging

Individually blister packaged

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service