Skip to Content
Merck
All Photos(1)

Documents

PAPP1-160K

Millipore

MILLIPLEX® Porcine Acute Phase Protein Panel 1

Synonym(s):

Luminex porcine acute phase multiplex kit, porcine multiplex CRP, porcine multiplex haptoglobin, Porcine multiplex kit, Luminex porcine immunoassay, Millipore porcine panel, porcine acute phase multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

porcine

Quality Level

manufacturer/tradename

Milliplex®

parameter

25 μL sample volume (per well of a 1:5000 dilution)

assay range

recovery range: 101%
(Haptoglobin)

recovery range: 102%
(CRP)

standard curve range: 0.15-600 ng/mL
(CRP)

standard curve range: 0.24-1000 ng/mL
(Haptoglobin)

inter-assay cv: ≤15%
intra-assay cv: ≤10%

technique(s)

multiplexing: suitable

input

sample type plasma
sample type serum

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Acute phase proteins (APPs) constitute an integral part of the innate immune response, exhibiting significant alterations in their serum/plasma concentrations in response to inflammation, infection, or trauma. This group of proteins includes haptoglobin (Hp) and C-reactive protein (CRP), which play pivotal roles in immune response, and are frequently studied as biomarkers of systemic inflammation and disease.
Research category: Cardiovascular
Research sub-category: Inflammation

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

MILLIPLEX® Porcine Acute Phase Protein Panel 1 is a 2-plex kit to be used for the simultaneous quantification of any or all of the following analytes in porcine serum and plasma samples: C-Reactive Protein (CRP) and Haptoglobin.

Features and Benefits

Configurable

Packaging

Each kit sufficient for 96 wells

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service