MXSMC0120
Milliflex Solid Media Cassette
Empty Milliflex Cassettes to be filled with solid media
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116129
NACRES:
NA.74
Recommended Products
material
polypropylene
styrene-acrylonitrile (SAN)
Quality Level
sterility
sterile
form
plate (solid agar)
feature
ready-to-use
manufacturer/tradename
Milliflex®
input
sample type bottled water
sample type cosmetics
sample type beer
sample type pharmaceutical(s)
application(s)
bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Milliflex® Cassette are empty plastic plates designed to be dispensed with agar-based media. It is to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The design of the plate helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly for dispensing 10ml of the media. The plate is sterilized and comes in 120 units per pack.
Application
Milliflex® empty Solid Media Cassette is used in Bioburden testing and general microbiological analysis of water. It is widely used in pharmaceutical, cosmetic and brewing industry
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service