MXRDP2KT01
Milliflex® Rapid System 2.0
for use with Milliflex Oasis®, suitable for bioburden testing, sterility testing
About This Item
Recommended Products
AC/DC input
100 V / 230 V, 50-60 Hz
Agency
EP 2.6.1
USP 71
71
product line
Milliflex®
packaging
pkg of 1 kit
parameter
10 kg operating weight (22 lb) (Autospray Station)
5 kg operating weight (11 lb) (Detection Tower)
technique(s)
Rapid Sterility Testing: suitable
H × W × D
31 cm (12 in.) × 21 cm (8 in.) × 40 cm (16 in.), Auto Spray Station
35 cm (14 in.) × 30 cm (12 in.) × 13 cm (5 in.), Detection Tower
application(s)
bioburden testing
sterility testing
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis®
General description
The detection method is based on highly sensitive ATP bioluminescence technology. The ATP method is regulatory compliant, highly reliable, and consistent with the detection rates matching the standards of current compendial testing methods.
The conventional way of visual evaluation is replaced by Complementary Metal Oxide Semiconductor (CMOS) in Milliflex® Rapid system. The state-of-the-art image analysis and optimized reagents require reduced incubation time for viable microbial detection.
The image analysis software intensifies (~1000 times) the bioluminescence from each microbial micro-colony and captures the light signals emitted by the microbes. The software enumerates the colonies and displays results as CFU (colony forming unit).
Application
Features and Benefits
- Consistent, fast, and reproducible results.
- Correlation of CFU counts with that of traditional methods.
- Data Integrity Software compliant with 21 CFR Part 11.
- Easy Handling and Validation.
Components
- Detection Tower -MXRDP2DT00)
- Auto Spray station - MXRP2SPRKT
- 2 x Power Supplies required (not included, sold separately, region specific under MXRPWRSP--)
Other Notes
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service