Skip to Content
Merck

Skip To

MX0490

Methanol

≥98% (GC), histological grade

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
UNSPSC Code:
41116121
EC Index Number:
200-659-6
MDL number:
Beilstein/REAXYS Number:
1098229
NACRES:
NA.21
Assay:
≥98% (GC)
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol,

vapor density

1.11 (vs air)

Quality Segment

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

assay

≥98% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

impurities

≤0.002% Acetone and Aldehydes, ≤0.5% Water (H2O)

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

histology

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Application

Used as a fixative for hematology and a substitute for ethanol.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Acetone and Aldehydes: 0.002% max
Assay (GC): 98% min
Water (H2O): 0.5% max
Evaporation Number: 1.9

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR13721.13351MX0487
application(s)

histology

application(s)

pharmaceutical (small molecule)

application(s)

food and beverages
pharmaceutical

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

assay

≥98% (GC)

assay

-

assay

≥99.8% (GC)

assay

≥99.8% (GC)

form

liquid

form

-

form

liquid

form

liquid

bp

64.7 °C (lit.)

bp

64.7 °C (lit.)

bp

64.7 °C (lit.)

bp

64.7 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters