Skip to Content
Merck

Skip To

MPTRB

Transfer buffer powder for use with mPAGE® Bis-Tris gels

pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X)

Synonym(s):

Transfer Buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

packaging

pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X)

General description

Transfer buffer powder for use with mPAGE Bis-Tris gels. 10 packets. Each packets makes 1 liter of transfer buffer at 1X.

  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

mPAGE Transfer Buffer is formulated fortransfer of proteins from mPAGE Bis-Tris gels to blotting membranes. For standard standard wet (tank) transfer, use the buffer at 1X. For semi-dry transfer, use the buffer at 2X. Please see the mPAGE user guide for instructions on reconstituting the buffer.

Western Blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MPMESMPM0PSMP10W15
form

powder

form

powder

form

powder

form

-

packaging

pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X)

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

packaging

pkg of 10 gels


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters