Skip to Content
Merck
All Photos(6)

Key Documents

MMP000864

Millipore

Milliflex® Oasis Funnel

Individually Packed, for use with Milliflex Oasis® systems, gridded black mixed cellulose esters (MCE) filter membrane, pore size 0.45 μm

Synonym(s):

Milliflex Oasis® filtration unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

material

gridded black mixed cellulose esters (MCE) filter membrane
polypropylene body
styrene-acrylonitrile (SAN) body

Quality Level

sterility

sterile; ethylene oxide treated

product line

Milliflex Oasis®

feature

ready-to-use

parameter

100 mL max. working volume

pore size

0.45 μm

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® systems

Looking for similar products? Visit Product Comparison Guide

General description

Milliflex® Oasis Funnel is a pre-sterilized, ready-to-use filtration unit. The unit is comprised of:
  • 100ml funnel.
  • Black gridded MCE filter membrane.

The sealed integral membrane filter eliminates the need for membrane manipulation and handling, thus minimizing a major source of contamination and thereby improves bioburden testing reliability. The sealed membrane prevents the bypass phenomenon often found with other filtration systems.
The Milliflex® filtration possesses almost twice the surface area of standard membranes enabling faster filtration and the funnel can be easily detached from the membrane without the use of any tools. The convex shape of the membrane after filtration demonstrates the integrity of the membrane and prevents air bubbles from being trapped when placed on culture media.

Application

Milliflex® Oasis Funnel is used during filtration of samples for Bioburden testing, water monitoring and general microbiological analysis of beer, bottled water. It is widely used in pharmaceutical, cosmetic, and brewing industry.

Features and Benefits

  • Effortless liquid level control, due to the fully transparent material
  • Limited liquid residues after filtration, due to the ultra-hydrophobic property of the material
  • Effortless funnel and membrane separation
  • Reduced risk of secondary contamination, due to the new hinged lid which allows the device to be closed during filtration

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service