Skip to Content
Merck
All Photos(1)

Key Documents

MDRPN0410

Millipore

Multiscreen® 96 well Plate, PTFE membrane

pore size 0.45 μm, non-sterile

Synonym(s):

96-Well PTFE Plate, PTFE 96-Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.24

material

PTFE membrane (hydrophobic)
flat bottom wells
polyolefin copolymer

Quality Level

description

Clear, Non-Sterile, 0.45 µm pore size, Hydrophobic Polytetrafluoroethylene (PTFE), 10 plates, volume 1.9mL

sterility

non-sterile

product line

MultiScreen®

feature

hydrophobic
lid: no

manufacturer/tradename

MultiScreen®

parameter

1.9 mL sample volume (per well)

H

40.7 mm

L

128 mm

W

85.5 mm

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

1.9 mL

working volume

1.8 mL

pore size

0.45 μm pore size

application(s)

sample preparation

binding type

low binding surface

shipped in

ambient

General description

The MultiScreen® Deep Well Solvinert Filter Plate is intended for a wide range of applications including total drug analysis, bead cleavage (solid-phase combinatorial chemistry and peptide synthesis), and sample preparation before high-performance liquid chromatography (HPLC)/ ultraviolet (UV) and liquid chromatography-tandem mass spectrometry (LC-MS/MS). This filter plate is fully automation compatible.

Application

MultiScreen® Deep, 96 Well Solvinert has been used to produce adsorption equilibrium isotherms. It has also been used in protein binding experiments.

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recovery of sinapic acid from canola/rapeseed meal extracts by adsorption
Moreno GM, et al.
Food and Bioproducts Processing, 120, 69-79 (2020)
Beckley K Nfor et al.
Journal of chromatography. A, 1217(44), 6829-6850 (2010-10-01)
The thermodynamic modeling of protein adsorption on mixed-mode adsorbents functionalized with ligands carrying both hydrophobic and electrostatic groups was undertaken. The developed mixed mode isotherm was fitted with protein adsorption data obtained for five different proteins on four different mixed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service