MCR4010FS1
Millistak+® Pod Depth Filter
CR40, filtration area 1.1 m², Process Scale
Synonym(s):
Millistak+ Pod Depth Filter, CR40 media series, 1.1 m 2 surface area, 1/pack
About This Item
Recommended Products
Product Name
Millistak+® Pod Depth Filter, CR40, cellulose, filtration area 1.1 m2, hydrophilic
material
carbon (active)
cellulose
polypropylene
Quality Level
description
Activated carbon is a very versatile process aid and has a high affinity and adsorption capacity for odorous compounds and color bodies over a wide range of molecular weights
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
1 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
H
32 cm (12.5 in.)
L
62 cm (24.2 in.)
filtration area
1.1 m2
impurities
<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
total organic carbon (TOC) residue
1300-1700 ppb
input
sample type liquid
conductivity
4.36-4.57 μS/cm at 25 °C (product water)
filter grade
CR40
Related Categories
General description
Application
Specifications
Preparation Note
Integrity is maintained after 1 autoclave cycle of 60 minutes @ 123 °C
Analysis Note
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service