Skip to Content
Merck
All Photos(2)

Key Documents

MABF297

Sigma-Aldrich

Anti-RIG-I, clone 1C3 Antibody

clone 1C3, from mouse

Synonym(s):

Probable ATP-dependent RNA helicase DDX58, DEAD box protein 58, RIG-I-like receptor 1, RLR-1, Retinoic acid-inducible gene 1 protein, RIG-1, Retinoic acid-inducible gene I protein, RIG-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C3, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDX58(23586)

General description

Retinoic acid-inducible gene 1 protein (RIG-1) is a cytoplasmic DExD/H box RNA helicase belonging to the helicase family and containing 2 CARD domains within the N-terminus, a repressor domain (RD), a helicase ATP-binding domain and a helicase C-terminal binding domain. RIG-1 is able to bind the 5’-triphosphate RNA of single or double-stranded viral RNA. After recognizing and binding viral RNA, RIG-1 protein initializes type I IFN production, and within fibroblasts and dendritic cells plays a central role in the regulation of type I IFN production in response to viral presence. There are four known isoforms resulting from alternative splice variants.

Immunogen

Recombinant protein corresponding to human RIG-I.

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated RIG-1 in A-549 infected with SeV cell lysate (Baum, A., et al. (2011). PNAS. 108(7):16303-16308).
Immunoprecipitation Analysis: A representative lot immunoprecipitated RIG-1 in A-459 lung carcinoma cell lysate (Nistal-Villan, E., et al. (2010). J. Biol. Chem. 285:20252-20261).
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected RIG-I in HeLa cells (Courtesy of Maite Sanchez).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-RIG-I, clone 1C3 Antibody is validated for use in Western Blotting and Immunoprecipitation and Immunocytochemistry for the detection of RIG-I.

Quality

Evaluated by Western Blotting in A-549 infected with SeV cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected RIG-1 in 10 µg of A-549 infected with SeV cell lysate.

Target description

~110 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1λ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippa Hillyer et al.
Journal of virology, 92(15) (2018-05-18)
Respiratory syncytial virus (RSV) infects small foci of respiratory epithelial cells via infected droplets. Infection induces expression of type I and III interferons (IFNs) and proinflammatory cytokines, the balance of which may restrict viral replication and affect disease severity. We
Na Li et al.
Cell reports, 35(6), 109091-109091 (2021-05-08)
It is urgent and important to understand the relationship of the widespread severe acute respiratory syndrome coronavirus clade 2 (SARS-CoV-2) with host immune response and study the underlining molecular mechanism. N6-methylation of adenosine (m6A) in RNA regulates many physiological and
Daniel T Thoresen et al.
Molecular cell, 83(1), 90-104 (2022-12-16)
RIG-I is essential for host defense against viral pathogens, as it triggers the release of type I interferons upon encounter with viral RNA molecules. In this study, we show that RIG-I is rapidly and efficiently activated by small quantities of
Jie Chen et al.
Cell reports, 32(9), 108080-108080 (2020-09-03)
The DNA-dependent pattern recognition receptor, cGAS (cyclic GMP-AMP synthase), mediates communication between the DNA damage and the immune responses. Mitotic chromosome missegregation stimulates cGAS activity; however, it is unclear whether progression through mitosis is required for cancercell-intrinsic activation of anti-tumor
Soham Gupta et al.
PLoS pathogens, 14(1), e1006852-e1006852 (2018-01-23)
The N-terminal domains of the herpesvirus large tegument proteins encode a conserved cysteine protease with ubiquitin- and NEDD8-specific deconjugase activity. The proteins are expressed during the productive virus cycle and are incorporated into infectious virus particles, being delivered to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service