Skip to Content
Merck
All Photos(1)

Key Documents

MABE61

Sigma-Aldrich

Anti-macro-H2A1.2 Antibody, clone 14G7

clone 14G7, from mouse

Synonym(s):

H2A histone family, member Y, Histone macroH2A1, Medulloblastoma antigen MU-MB-50.205, histone macroH2A1.1, histone macroH2A1.2, Core histone macro-H2A.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14G7, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2AFJ(55766)

General description

Histone MacroH2A (mH2A), also known as H2AFY, is a vertebrate-specific histone variant associated with condensed, inactive heterochromatin. This histone variant consists of a histone H2A-like domain fused to a large non-histone region. mH2A has been shown to repress RNA polymerase II-driven transcription at the level of transcriptional initiation. The non-histone region of mH2A is directly involved in the repression of transcription, histone acetylation and blocking SWI/SNF and ACF nucleosome remodeling. This variant histone is enriched in chromatin of the female inactive X chromosome and in highly methylated CpG loci.

Immunogen

GST-tagged recombinant protein corresponding to human macro-H2A1.2.

Application

Anti-macro-H2A1.2 Antibody, clone 14G7 is a Mouse Monoclonal Antibody for detection of macro-H2A1.2 also known as H2A histone family member Y or Histone macroH2A1 & has been validated in WB, ICC & IHC.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH. (Sporn, J.C., et al. (2009). Oncogene. 1–6.)

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in IC. (Sporn, J.C., et al. (2009). Oncogene. 1–6.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in MCF-7 cell lysate.

Western Blot Analysis: 0.25 µg/mL of this antibody detected macro-H2A1.2 on 10 µg of MCF-7 cell lysate.

Target description

~39 kDa observed

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2bλ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF-7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murali Palangat et al.
Genes & development, 33(9-10), 482-497 (2019-03-08)
Somatic mutations in the genes encoding components of the spliceosome occur frequently in human neoplasms, including myeloid dysplasias and leukemias, and less often in solid tumors. One of the affected factors, U2AF1, is involved in splice site selection, and the
Simran Khurana et al.
mBio, 12(6), e0268421-e0268421 (2021-11-10)
The life cycle of human papillomavirus (HPV) depends on keratinocyte differentiation as the virus modulates and takes advantage of cellular pathways to replicate its genome and assemble viral particles in differentiated cells. Viral genomes are amplified in nuclear replication foci
Florian Wimmers et al.
Cell, 184(15), 3915-3935 (2021-06-27)
Emerging evidence indicates a fundamental role for the epigenome in immunity. Here, we mapped the epigenomic and transcriptional landscape of immunity to influenza vaccination in humans at the single-cell level. Vaccination against seasonal influenza induced persistently diminished H3K27ac in monocytes
Jeongkyu Kim et al.
Molecular cell, 69(1), 36-47 (2017-12-19)
Recent integrative epigenome analyses highlight the importance of functionally distinct chromatin states for accurate cell function. How these states are established and maintained is a matter of intense investigation. Here, we present evidence for DNA damage as an unexpected means
Ramhari Kumbhar et al.
The Journal of cell biology, 220(7) (2021-05-19)
The histone demethylase KDM5A erases histone H3 lysine 4 methylation, which is involved in transcription and DNA damage responses (DDRs). While DDR functions of KDM5A have been identified, how KDM5A recognizes DNA lesion sites within chromatin is unknown. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service