Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
NCM II 100, monoclonal
Application:
ICC, IP, WB
Citations:
5
biological source
mouse
Quality Segment
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
NCM II 100, monoclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MYCN(4613)
General description
The v-myc oncogene, initially identified in the MC29 avian retrovirus, causes myelocytomas, carcinomas, sarcomas and lymphomas, and belongs to a family of oncogenes conserved throughout evolution. In humans the family consists of five genes: c-myc, N-myc, R-myc, L-myc and B-myc. Amplification of the N-myc gene has been found in human neuroblastomas and cell lines. The extent of N-myc amplification correlates well with the stage of neuroblastoma disease. Immunological studies have shown that the human N-myc gene gives rise to at least two nuclear phosphoproteins that exhibit relatively short (30 min) half lives in vivo and exhibit DNA binding properties in vitro.
~55 kDa observed. Uniprot describes a molecular weight at ~50 kDa Uniprot states that this protein may be phosphorylated.
Immunogen
Recombinant protein corresponding to human N-Myc.
Application
Immunofluorescence Analysis: A representative lot from an independent laboratory detected N-Myc in IMR5 cells (Ikegaki, N., et al. (1986). 83(16):5929-5933.).
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated N-Myc in IP (Brondyk, W. H., et al. (1991). 6(7):1269-1276.).
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated N-Myc in IP (Brondyk, W. H., et al. (1991). 6(7):1269-1276.).
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Transcription Factors
Use Anti-N-Myc Antibody, clone NCM II 100 (mouse monoclonal antibody) validated in WB, IP, ICC to detect N-Myc also known as N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, bHLHe37.
Physical form
Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
IMR-32 cell lysate
IMR-32 cell lysate
Evaluated by Western Blot in IMR-32 cell lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected N-Myc in 10 µg of IMR-32 cell lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected N-Myc in 10 µg of IMR-32 cell lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.