Skip to Content
Merck
All Photos(1)

Key Documents

MAB9013

Sigma-Aldrich

Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D

clone 542-7D, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

542-7D, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

Coronavirus nucleoprotein; Strain OC-43

Immunogen

Epitope: nucleoprotein of OC-43.

Application

Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D detects level of Coronavirus Group Antigen & has been published & validated for use in IF.
IFA. Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified Immunoglobulin. Supplied in PBS buffer pH 7.4. Contains 0.1% sodium azide and carrier protein.

Storage and Stability

Maintain at +4°C for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jang-Hoon Choi et al.
PloS one, 15(5), e0232757-e0232757 (2020-05-10)
Middle East respiratory syndrome coronavirus (MERS-CoV) causes severe respiratory infection and continues to infect humans, thereby contributing to a high mortality rate (34.3% in 2019). In the absence of an available licensed vaccine and antiviral agent, therapeutic human antibodies have
Mario Milani et al.
Antiviral research, 189, 105055-105055 (2021-03-14)
The current emergency of the novel coronavirus SARS-CoV2 urged the need for broad-spectrum antiviral drugs as the first line of treatment. Coronaviruses are a large family of viruses that already challenged humanity in at least two other previous outbreaks and
Kentaro Uemura et al.
iScience, 24(10), 103120-103120 (2021-09-21)
Newly emerging or re-emerging viral infections continue to cause significant morbidity and mortality every year worldwide, resulting in serious effects on both health and the global economy. Despite significant drug discovery research against dengue viruses (DENVs) and severe acute respiratory
Gergely Rona et al.
Cell death and differentiation, 29(2), 285-292 (2021-12-05)
The risk of zoonotic coronavirus spillover into the human population, as highlighted by the SARS-CoV-2 pandemic, demands the development of pan-coronavirus antivirals. The efficacy of existing antiviral ribonucleoside/ribonucleotide analogs, such as remdesivir, is decreased by the viral proofreading exonuclease NSP14-NSP10
Daniel Poston et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 73(5), e1208-e1211 (2020-12-04)
Cross-reactive immune responses elicited by seasonal coronaviruses might affect severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) susceptibility and disease outcomes. We measured neutralizing activity against SARS-CoV-2 in prepandemic sera from patients with prior polymerase chain reaction scan-confirmed seasonal coronavirus infection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service