Skip to Content
Merck
All Photos(1)

Key Documents

MAB8754

Sigma-Aldrich

Anti-Eastern Equine Encephalitis Antibody, Virus, clone 1A4B.6

clone 1A4B.6, Chemicon®, from mouse

Synonym(s):

EEE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

1A4B.6, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
inhibition assay: suitable (hemagglutination)

isotype

IgG2b

shipped in

wet ice

Specificity

The MAb 1A4B-6 is an IgG2b antibody that reacts with an
E1 epitope shared by all alphaviruses. Encephalitis Virus: Strains Indentified:

Eastern Equine (EEE) N. American eastern equine (NJ/60);

S. American eastern equine (BeAn 5122)

Western Equine (WEE) Western equine (McMIllan); Highlands J (B-230);

Sindbis

Semliki Forest Semliki Forest

Venezuelan Equine (VEE) 1AB (TC-83 Vaccine, Trinidad Donkey, PTF-39);

1C (P676); 1D (3880); 1E ( Mena II);

1F(78V-3531); 2 (Fe3-7c); 3 (Mucambo);

4 (Pixuna); 5 (Cabassou)

Immunogen

Epitope: Virus

Application

Detect the Eastern Equine Encephalitis protein using this Anti-Eastern Equine Encephalitis, Virus, clone 1A4B.6 validated for use in ELISA, IF & Hemagglutination Inhibition.
ELISA

Indirect Immunofluorescence

Hemagglutination Inhibition

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS with 0.1% Sodium Azide.

Storage and Stability

Maintain at 2 to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel Kruchelski Tschá et al.
Virus evolution, 7(2), veab105-veab105 (2022-03-22)
Genomic and epidemiological surveillance are paramount for the discovery of new viruses with the potential to cross species barriers. Here, we present a new member of the genus Alphavirus found in Trichoprosopon and Wyeomia mosquitoes, tentatively named Pirahy virus (PIRAV).
Brandon J Beddingfield et al.
NPJ vaccines, 9(1), 47-47 (2024-02-28)
MVA-based monovalent eastern equine encephalitis virus (MVA-BN-EEEV) and multivalent western, eastern, and Venezuelan equine encephalitis virus (MVA-BN-WEV) vaccines were evaluated in the cynomolgus macaque aerosol model of EEEV infection. Macaques vaccinated with two doses of 5 × 108 infectious units
Efficient functional pseudotyping of oncoretroviral and lentiviral vectors by Venezuelan equine encephalitis virus envelope proteins
Kolokoltsov, Andrey A, et al
Journal of virology, 79, 756-763 (2005)
Detection of eastern equine encephalitis virus in infected mosquitoes using a monoclonal antibody-based antigen-capture enzyme-linked immunosorbent assay
Brown, T M, et al
The American Journal of Tropical Medicine and Hygiene, 65, 208-213 (2001)
Matthew S Sutton et al.
Cell, 186(12), 2672-2689 (2023-06-10)
Alphaviruses are RNA viruses that represent emerging public health threats. To identify protective antibodies, we immunized macaques with a mixture of western, eastern, and Venezuelan equine encephalitis virus-like particles (VLPs), a regimen that protects against aerosol challenge with all three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service