Skip to Content
Merck
All Photos(1)

Documents

MAB4315

Sigma-Aldrich

Anti-STRO-1 Antibody, clone STRO-1

ascites fluid, clone STRO-1, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

STRO-1, monoclonal

species reactivity

human, primate

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

input

sample type mesenchymal stem cell(s)

isotype

IgM

shipped in

dry ice

target post-translational modification

unmodified

General description

STRO-1 is a cell surface protein expressed by bone marrow stromal cells and erythroid precursors. The frequency of colony forming units fibroblasts (CFU-F) was enriched 100-fold in the STRO-1+/Glycophorin A- population from bone marrow cells. The subset of marrow cells that expresses the STRO-1 antigen is capable of differentiating into multiple mesenchymal lineages including hematopoiesis-supportive stromal cells with a vascular smooth muscle-like phenotype, adipocytes, osteoblasts, and chondrocytes.

Specificity

Recognizes a cell surface antigen expressed by bone marrow stromal cells and stromal precursors. STRO-1 binds to approximately 10% of bone marrow mononuclear cells, greater than 95% of which are nucleated erythroid precursors.

Immunogen

CD34+ bone marrow cells

Application

Detect STRO-1 using this Anti-STRO-1 Antibody, clone STRO-1 validated for use in FC & IF.

Analysis Note

Control
Bone marrow cultures

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung Cheol Kim et al.
Journal of biomedical research, 33(2), 122-130 (2019-04-24)
Despite their potential applications in future regenerative medicine, periodontal ligament stem cells (PDLSCs) are difficult to obtain in large amounts from patients. Therefore, maintaining stemness while expanding the cell numbers for medical use is the key to transitioning PDLSCs from
Liuhua Zhou et al.
PloS one, 10(2), e0117644-e0117644 (2015-02-24)
Autologous endothelial progenitor cells (EPCs) might be alternative angiogenic cell sources for vascularization of tissue-engineered bladder, while isolation and culture of EPCs from peripheral blood in adult are usually time-consuming and highly inefficient. Recent evidence has shown that EPCs also
Yiping Fan et al.
PloS one, 5(12), e14421-e14421 (2011-01-05)
Mammary stem cells have been extensively studied as a system to delineate the pathogenesis and treatment of breast cancer. However, research on mammary stem cells requires tissue biopsies which limit the quantity of samples available. We have previously identified putative
Srinivas Velugotla et al.
Biomicrofluidics, 6(4), 44113-44113 (2012-01-01)
Assessment of the dielectrophoresis (DEP) cross-over frequency (f xo), cell diameter, and derivative membrane capacitance (C m) values for a group of undifferentiated human embryonic stem cell (hESC) lines (H1, H9, RCM1, RH1), and for a transgenic subclone of H1
Jeong Seok Lee et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 34(5), 1824-1834 (2014-12-17)
Mussel-inspired polydopamine (PDA) is known to be an effective bioadhesive and bioactive material for controlling stem cell fate, which is important in stem cell-based regenerative medicine; however, the effect of PDA on osteogenic differentiation of periodontal ligament stem cells (PDLSCs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service