Skip to Content
Merck
All Photos(1)

Key Documents

MAB3484

Sigma-Aldrich

Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, a.a. 386-412, clone L12B4

clone L12B4, Chemicon®, from mouse

Synonym(s):

CFTR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L12B4, monoclonal

species reactivity

human

should not react with

shark, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CFTR(1080)

Specificity

Human cystic fibrosis transmembrane conductance regulator (CFTR). Recognizes an epitope at the cytoplasmic region preceding the first nucleotide binding domain between residues 386 to 412.

Recognizes CFTR, Mr 170kDa and two additional proteins at ~ Mr 100 and 140kDa.

Immunogen

Epitope: a.a. 386-412 of human CFTR

Application

Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, a.a. 386-412, clone L12B4 is an antibody against Cystic Fibrosis Transmembrane Conductance Regulator for use in IC, IP & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10μg/mL, note do not boil the lysate, incubate at 80C for 30 minutes prior to running SDS-PAGE. CTFR aggregates upon boiling. Antibody recognizes CFTR at 170kDa and two additional proteins at 100 & 140kDa.

Immunoprecipitation

Immunofluorescence

Note: Does not work on paraffin embedded tissue.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graeme W Carlile et al.
Scientific reports, 12(1), 4595-4595 (2022-03-19)
Most cases of cystic fibrosis (CF) are caused by class 2 mutations in the cystic fibrosis transmembrane regulator (CFTR). These proteins preserve some channel function but are retained in the endoplasmic reticulum (ER). Partial rescue of the most common CFTR
Chiara Brandas et al.
Biomolecules, 11(10) (2021-10-24)
Cystic fibrosis (CF) is caused by loss-of-function mutations in the CF transmembrane conductance regulator (CFTR) protein, an anion channel that regulates epithelial surface fluid secretion. The deletion of phenylalanine at position 508 (F508del) is the most common CFTR mutation. F508del
Celeste Riepe et al.
Molecular biology of the cell, 35(2), ar15-ar15 (2023-11-29)
Over 80% of people with cystic fibrosis (CF) carry the F508del mutation in the cystic fibrosis transmembrane conductance regulator (CFTR), a chloride ion channel at the apical plasma membrane (PM) of epithelial cells. F508del impairs CFTR folding causing it to
Mauricio Di Fulvio et al.
PloS one, 15(12), e0242749-e0242749 (2020-12-03)
Cystic fibrosis (CF) is due to mutations in the CF-transmembrane conductance regulator (CFTR) and CF-related diabetes (CFRD) is its most common co-morbidity, affecting ~50% of all CF patients, significantly influencing pulmonary function and longevity. Yet, the complex pathogenesis of CFRD
Javier Sanz et al.
European journal of human genetics : EJHG, 18(2), 212-217 (2009-09-03)
Cystic fibrosis (CF) is one of the most common genetic diseases in the Caucasian population and is characterized by chronic obstructive pulmonary disease, exocrine pancreatic insufficiency, and elevation of sodium and chloride concentrations in the sweat and infertility in men.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service