Skip to Content
Merck
All Photos(1)

Key Documents

MAB3391

Sigma-Aldrich

Anti-Collagen Type I (COL1A1) Antibody

CHEMICON®, mouse monoclonal, 5D8-G9

Synonym(s):

Anti-CAFYD, Anti-EDSARTH1, Anti-EDSC, Anti-OI1, Anti-OI2, Anti-OI3, Anti-OI4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Collagen Type I Antibody, clone 5D8-G9, clone 5D8-G9, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D8-G9, monoclonal

species reactivity

pig, canine, sheep, human, bovine

should not react with

guinea pig, ground squirrel, rat, kangaroo, horse, chicken, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

Related Categories

General description

Collagens are highly conserved throughout evolution and are characterised by an uninterrupted "Glycine X Y" triplet repeat that is a necessary part of the triple helical structure. Type I collagen (95 kDa) is found in bone, cornea, skin and tendon. Mutations in the encoding gene are associated with osteogenesis imperfecta, Ehlers Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for Platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor.

Specificity

Collagen Type I Antibody is a monoclonal IgG1 antibody which binds an epitope near the C-terminal of Type I Collagen.

Collagen Type I Antibody displays a high affinity for human, bovine and ovine Type I Collagens. There is no evidence for cross reactivity with Collagen Types III, V and VI or connective tissue protein. Antibody reacts only with native, non-denatured Collagen I

Application

ELISA: detection of native type I collagen. Capture: 2-4 μg/ml; Detection: A dilution of 10μg/mL of Collagen Type I Antibody will detect 50ng of human Collagen Type I with no reactivity with other Collagens. Higher levels of antibody may be necessary to detect lower concentrations of Collagen. Note antibody will not pair with itself.

Immunohistochemistry:unfixed frozen sections:

Cut 6 μm thick sections from frozen samples using a freezing microtome. Air-dry throughly. Rehydrate with PBS. Blocking is usually not necessary for IF studies, if using DAB, pretreatment for peroxidase is suggested and blocking 2% BSA in PBS will reduce back ground.

Dilute Collagen Type I Antibody in PBS buffer, add to the sample(s) and incubate for 60 minutes. (1:100-1:500).

Wash sections for 10 minutes in PBS, twice. Add FITC conjugated anti-mouse antibody and incubate for 60 minutes.

Wash for 10 minutes in PBS, twice.

Mount sections in glycerol/water (9:1 v/v) containing 1 mM 1,4-phenylenediamine. (for FITC stability or use Chemicon catalog numbers 5096 or 5013.

NOTE:

Preliminary studies suggest that enhanced staining of certain tissues maybe obtained by pretreatment (previous to step 2) of sections with various enzymes (eg 0.1% pepsin in 0.1M HCl, 37°C for 5 minutes).

Flow Cytometry: 1:100

Immunoblotting:1:1000 using NATIVE, non-denatured, non-reduced western blots only. Ramshaw, et.al (1988) "Electrophoretic and electroblotting of native collagens." Anal. Biochem. 168:82-87. Antibody will not work in traditional, reduced western blots.

Briefly, PAGE gels must be prepared for running native collagens.

A 3% (w/v) total acrylamide separating gel, containing 3.2% (w/w) bis-acrylamide as a proportion to a monomer as the crosslinking agent, in 10mM calcium lactate, pH 6.8 is polymerized between vertical glass plates by the addition of 0.05-0.1% TEMED and 0.05-.1% ammonium persulfate (added from a 10% stock solution). The cathode buffer (negative) buffer is 50mM Tris adjusted to pH 6.6 with lactic acid; the anode (positive) buffer is 0.1M lactic acid pH 2.5 {Friis, SJ et al, 1985 J Biochem Biophys Methods 10:301-306.}.

Prior to sample loading the gel is run for at least 90 minutes at 100V. Collagen samples are dissolved at 1mg/mL in either 0.1M lactic acid or 0.1M acetic acid containing 10% sucrose, and 5-10μg per lane is loaded. Methyl green is added as required to assist loading. Electrophoresis is 70V for 5 hours, room temperature.

Blotting uses 0.1M lactic acid pH 2.5, and 20V for 16 hours at room temperature with the collagens migrating toward the cathode.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Collagen Type I Antibody, clone 5D8-G9 is validated for use in ELISA, FC, WB, IH for the detection of Collagen Type I.

Physical form

100μg purified antibody at a concentration of 1mg/mL in 50 mM Tris Acetate, 150 mM NaCl.

The immunoglobulin fraction has been purified by Protein A Chromatography and shown to be >95% pure by coomassie PAGE. The Collagen Type I Antibody is supplied 0.22 micron filtered.
Format: Purified

Storage and Stability

Collagen Type I Antibody is shipped, in liquid form, at ambient temperature. This material is stable for up to 6 months when stored at 2-8°C.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Analysis Note

Control
Human kidney lysate

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Marmotti et al.
European cells & materials, 26, 15-31 (2013-08-06)
We propose a culture-free approach to osteochondral repair with minced autologous cartilage fragments loaded onto a scaffold composed of a hyaluronic acid (HA)-derived membrane, platelet-rich fibrin matrix (PRFM) and fibrin glue. The aim of the study was to demonstrate in
M A Zarog et al.
BJS open (2020-10-14)
Improved diagnostic biomarkers are required for acute appendicitis. The circulating fibrocyte percentage (CFP) is increased in inflammatory states, but has not been studied in acute appendicitis. This study aimed to determine CFP in acute appendicitis and compare diagnostic accuracy with
Jie Zhu et al.
International journal of molecular sciences, 21(11) (2020-06-18)
Collagen type I is a major constituent of animal bodies. It is found in large quantities in tendon, bone, skin, cartilage, blood vessels, bronchi, and the lung interstitium. It is also produced and accumulates in large amounts in response to
Woojin M Han et al.
Nature materials, 15(4), 477-484 (2016-01-05)
Treatment strategies to address pathologies of fibrocartilaginous tissue are in part limited by an incomplete understanding of structure-function relationships in these load-bearing tissues. There is therefore a pressing need to develop micro-engineered tissue platforms that can recreate the highly inhomogeneous
Oxygen tension is a determinant of the matrix-forming phenotype of cultured human meniscal fibrochondrocytes.
Adesida, AB; Mulet-Sierra, A; Laouar, L; Jomha, NM
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service