Skip to Content
Merck
All Photos(1)

Key Documents

MAB2078Z

Sigma-Aldrich

Anti-Integrin α9β1 Antibody, clone Y9A2, azide free

clone Y9A2, Chemicon®, from mouse

Synonym(s):

MAB2078

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Y9A2, monoclonal

species reactivity

human, guinea pig

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

suitability

not suitable for Western blot
not suitable for immunohistochemistry

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA9(3680)

Specificity

Reactive with human and guinea pig alpha9beta1.

Immunogen

Murine L cells transfected with human alpha9.

Application

Detect Integrin α9β1 using this Anti-Integrin α9β1 Antibody, clone Y9A2, azide free validated for use in FC & IP.
Immunoprecipitation

FACS Analysis

Function blocking: 100% inhibition of alpha9beta1-mediated adhesion to the third fibronectin type III repeat in tenascin at <10 μg/mL.

Not effective for immunohistochemistry, immunocytochemistry or western blot.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with no preservatives.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Integrin alpha9beta1 positive tissues include basal keratinocytes, hepatocytes, muscle (smooth and skeletal), and airway epithelium.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Timoshenko et al.
British journal of cancer, 97(8), 1090-1098 (2007-10-04)
Vascular endothelial growth factor C (VEGF-C) is a lymphangiogenic factor over-expressed in highly metastatic, cyclooxygenase (COX)-2 expressing breast cancer cells. We tested the hypothesis that tumour-derived VEGF-C may play an autocrine role in metastasis by promoting cellular motility through one
Patrick M Lelliott et al.
ImmunoHorizons, 6(2), 170-183 (2022-02-24)
Anti-neutrophil cytoplasmic Ab (ANCA)-associated vasculitis (AAV) is a life-threatening condition characterized by improper activation of neutrophils and the release of neutrophil extracellular traps (NETs) in small vessels. This study aimed to explain the role of NETs in AAV pathogenesis by
Whitney M Longmate et al.
The Journal of cell biology, 216(5), 1473-1488 (2017-04-19)
Development of wound therapies is hindered by poor understanding of combinatorial integrin function in the epidermis. In this study, we generated mice with epidermis-specific deletion of α3β1, α9β1, or both integrins as well as keratinocyte lines expressing these integrin combinations.
Koichi Nishino et al.
Scientific reports, 11(1), 8406-8406 (2021-04-18)
Lymphangioleiomyomatosis (LAM) is a rare pulmonary disease characterised by the proliferation of smooth muscle-like cells (LAM cells), and an abundance of lymphatic vessels in LAM lesions. Studies reported that vascular endothelial growth factor-D (VEGF-D) secreted by LAM cells contributes to
Peggy Lafuste et al.
Molecular biology of the cell, 16(2), 861-870 (2004-12-03)
Knowledge on molecular systems involved in myogenic precursor cell (mpc) fusion into myotubes is fragmentary. Previous studies have implicated the a disintegrin and metalloproteinase (ADAM) family in most mammalian cell fusion processes. ADAM12 is likely involved in fusion of murine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service