Skip to Content
Merck
All Photos(1)

Key Documents

MAB1987

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

CHEMICON®, mouse monoclonal, P4C10

Synonym(s):

CD29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin β1 Antibody, clone P4C10, culture supernatant, clone P4C10, Chemicon®

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

P4C10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

Specificity

Reacts with the beta-1 subunit of the integrin protein family. Appears to be human specific. Unlike many beta-1 monoclonals 4C10′s epitope is not trypsin sensitive.

Application

Detect Integrin β1 using this Anti-Integrin β1 Antibody, clone P4C10 validated for use in ELISA, IP, WB & IC.
Inhibits attachment of UCLA-P3 cells to collagen type I (Wayner, 1991). Inhibits endodermal and keratinocytes to collagen, fibronectin and laminin. It also inhibits cell to cell adhesion. Typical dilution ranges are 1:50-1:500.

Immunoflourescence

Immunoblotting

Immunoprecipitation

ELISA

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Integrin β1 in HeLa cells.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Tissue Culture supernatant, containing 0.1% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fornaro et al.
Molecular biology of the cell, 11(7), 2235-2249 (2000-07-11)
The integrin cytoplasmic domain modulates cell proliferation, adhesion, migration, and intracellular signaling. The beta(1) integrin subunits, beta(1C) and beta(1A), that contain variant cytoplasmic domains differentially affect cell proliferation; beta(1C) inhibits proliferation, whereas beta(1A) promotes it. We investigated the ability of
W-C Liao et al.
Placenta, 36(4), 357-364 (2015-02-11)
Glycosylation controls diverse protein functions and regulates various cellular phenotypes. Trophoblast invasion is essential for normal placental development. However, the role of glycosylation in human placenta throughout pregnancy is still unclear. The β-1,4-galactosyltransferase III (B4GALT3) has been found to regulate
Platelet matrix metalloprotease-1 mediates thrombogenesis by activating PAR1 at a cryptic ligand site.
Trivedi, V; Boire, A; Tchernychev, B; Kaneider, NC; Leger, AJ; O'Callaghan, K; Covic et al.
Cell null
Tsutomu Sato et al.
Cancer research, 65(15), 6950-6956 (2005-08-03)
CD26 is an antigen with key role in T-cell biology and is expressed on selected subsets of aggressive T-cell malignancies. To elucidate the role of CD26 in tumor behavior, we examine the effect of CD26 depletion by small interfering RNA
Ann-Marie Baker et al.
Journal of the National Cancer Institute, 103(5), 407-424 (2011-02-02)
Emerging evidence implicates lysyl oxidase (LOX), an extracellular matrix-modifying enzyme, in promoting metastasis of solid tumors. We investigated whether LOX plays an important role in the metastasis of colorectal cancer (CRC). We analyzed LOX expression in a patient CRC tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service