Skip to Content
Merck
All Photos(1)

Documents

MAB1330

Sigma-Aldrich

Anti-Collagen Type II Antibody, clone COLL-II

ascites fluid, clone COLL-II, Chemicon®

Synonym(s):

Anti-ANFH, Anti-AOM, Anti-COL11A3, Anti-SEDC, Anti-STL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

COLL-II, monoclonal

species reactivity

sheep, human, bovine, rat, pig, mouse, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL2A1(1280)

Specificity

Reacts with both pepsin-solubilized and CNBr-cleaved human and bovine collagen type II. No cross-reactivity with collagen types I, III, V and IX.

Immunogen

Human (cartilage specific) CNBr-cleaved collagen II.

Application

Immunofluorescence: 1:2-1:5 with fresh, frozen or acetone-fixed material. Mild pepsin digestion is recommended to enhance staining.

Western Blot: 1:1000-1:2000 with chemiluminescent detection

ELISA at 1:100-1:500, pepsin treated samples.





Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Collagen Type II Antibody, clone COLL-II is validated for use in ELISA, IH for the detection of Collagen Type II.

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Cartilage in lung or fetus

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biwei Deng et al.
International journal of nanomedicine, 13, 8217-8230 (2018-12-18)
The osteoinductive behaviors of nitinol (NiTi)-based metal implants for bone regeneration are largely dependent on their surface composition and topology. Continuous-mode laser sintering often results in complete melting of the materials and aggregation of particles, which lack control of heat
Feng-Cheng Liu et al.
Nutrients, 11(5) (2019-05-30)
Genistein is an isoflavone extracted from soybean (Glycine max). This compound has anti-inflammatory, anti-oxidative, and anti-cancer effects; however, the mechanism underlying the effects of genistein on IL-1β-stimulated human osteoarthritis (OA) chondrocytes remains unknown. Our objectives in this study were to
L A Vonk et al.
Osteoarthritis and cartilage, 22(1), 145-153 (2013-11-26)
Hsa-miR-148a expression is decreased in Osteoarthritis (OA) cartilage, but its functional role in cartilage has never been studied. Therefore, our aim was to investigate the effects of overexpressing hsa-miR-148a on cartilage metabolism of OA chondrocytes. OA chondrocytes were transfected with
Xin Lu et al.
International journal of molecular medicine, 37(2), 509-516 (2015-12-29)
The aim of the present study was to examine the expression levels and role of hsa-miR-15a in osteoarthritis (OA), as well as the associated mechanisms. The expression levels of hsa-miR-15a and A disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type
Yang-Jun Lao et al.
Orthopaedic surgery, 10(1), 56-63 (2018-02-13)
To investigate the effect of accumulated spinal axial biomechanical loading on mice lumbar disc and the feasibility of applying this method to establish a mice intervertebral disc degeneration model using a custom-made hot plate cage. In previous studies, we observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service