Skip to Content
Merck
All Photos(4)

Documents

MAB1217F

Sigma-Aldrich

Anti-FMC-7 B-Cell Lymphocyte Marker Antibody, clone FMC-7, FITC conjugated

clone FMC-7, Chemicon®, from mouse

Synonym(s):

Anti-FMC-7, B-Cell Marker Detection, FITC Anti-FMC-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

clone

FMC-7, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

General description

B cells are lymphocytes that play a large role in the humoral immune response as opposed to the cell-mediated immune response that is governed by T cells. The principal function of B cells is to make antibodies against soluble antigens. B cells are an essential component of the adaptive immune system. FMC-7 B-Cell Lymphocyte Marker is a 105 kDa B cell restricted antigen which is expressed on about 50% of adult human peripheral blood B cells. Upon in vivo B cell activation FMC7 expression initially increases. B cells involved in antibody secretion have lost the FMC7 determinant. The monoclonal antibody FMC7 delineates a subpopulation of B lymphocytes in normal blood. Expression of the antigen recognized by FMC7 appears to be maturation-linked, and it serves to distinguish different types of B cell leukemia.
BIOCHEMISTRY: p I = 7.4

Specificity

The FMC-7 antibody was not clustered at the 5th Leucocyte Typing Workshop held in Boston USA 1993. This antibody detects a glycoprotein of 105kD found on circulating B-lymphocytes (Brooks et al., 1981). Studies on normal lymphocytes, leukaemic cells and cell lines indicate that FMC-7 is a marker for a limited segment of the B cell maturation pathway (Catovsky et al., 1981; Zola et al., 1984). This glycoprotein is expressed in varying degrees on normal circulating B-cells, chronic B-cell leukaemia (B-CLL), prolymphocytic leukaemia (PLL) and other B cell neoplasias (Zola et al., 1984, 1987; Bloem et al., 1988; Melo et al., 1988; Ferro & Zola, 1990; Mulligan, 1990; Zola et al., 1991; Burthem & Cawley, 1994; Huh et al., 1994). It labels the same population as CD22 antibodies but reacts with a different antigen (Zola et al. 1987).

Cell reactivity:1

Stains peripheral blood B lymphocytes and tonsil B lymphocytes. No reaction with granulocytes, monocytes, platelets, erythrocytes, T lymphocytes or null cells. Reacts with HRIK and Raji cell lines.

Clinical9,12,13 Expression

B cell prolymphocytic leukemia (B-PLL) Strongly positive

Hairy cell leukemia (HCL) Strongly positive

Hairy cell leukaemia variant (HCL-V) Strongly positive

Splenic lymphoma with villous lymphocytes (SLVL) Positive

B cell chronic lymphocytic leukaemia (B-CLL) Negative to weakly positive

Immunogen

The human B-lymphoblastoid cell line, HRIK.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-FMC-7 B-Cell Lymphocyte Marker Antibody, clone FMC-7, FITC conjugated is validated for use in Flow Cytometry (FC) for the detection of FMC-7 B-Cell Lymphocyte Marker.
This antibody can be used for the differentiation of PLL from B-CLL. It is also useful in confirmation of the diagnosis of other disorders such as HCL, HCL-V and SLVL. Useful for the detection of residual disease in patients undergoing treatment for these disorders.


SUGGESTED USAGE:

Flow cytometry - use 10 μl direct from the vial per 100 μl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.

Target description

105 kDa

Linkage

Replaces: CBL1416F

Physical form

The antibody is supplied in 1.0 ml 0.1M Tris-HCl, 0.5M NaCl, 10mM glycine, pH 8.0, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8°C, for up to 6 months. DO NOT FREEZE. Protect from light.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Analysis Note

Control
Human peripheral blood lymphocytes, HRIK and Raji cell lines

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service