Skip to Content
Merck
All Photos(1)

Documents

HEXSM-170K

Millipore

MILLIPLEX® Human Exosome Characterization Panel

Synonym(s):

EV multiplex kit, Human exosome multiplex kit, Luminex EV panel, Luminex human exosome immunoassay, Millipore human exosome panel, extracellular vesicle multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.55

species reactivity

human

Quality Level

packaging

pkg of 96 wells

manufacturer/tradename

Milliplex®

parameter

25 μL sample volume (per well)

assay range

intra-assay cv: ≤20%
standard curve range: 0.4-1,500 ng/mL
(CD9)

standard curve range: 0.5-2,000 ng/mL
(CD81)

standard curve range: 0.6-2,500 ng/mL
(Flotillin-1)

standard curve range: 0.9-3,500 ng/mL
(Syntenin-1)

standard curve range: 122-500,000 pg/mL
(CD63)

standard curve range: 24-100,000 pg/mL
(Calreticulin, GAPDH)

standard curve range: 49-200,000 pg/mL
(TSG101)

inter-assay cv: ≤10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Cells release extracellular vesicles (EV), including exosomes, as part of normal physiological processes including removal of waste and regulation of intercellular communication. Furthermore, increased exosome release has been implicated in the progression of various disease states and other abnormalities such as cancer and neurodegeneration. Exosome identification, classification, and differentiation from other extracellular vesicles is often complicated due to the variability in isolation and preparation protocols, exosome biogenesis, as well as exosome source. Thus, the development of additional exosome characterization tools is beneficial to the exosome research field. Commonly associated exosome-enriched markers include tetraspanins CD9, CD63, and CD81, cargo sorting-associated markers TSG101 and Flotillin-1, and biogenesis marker syntenin-1. Other markers, including Ago2, GAPDH, and Calreticulin, may or may not be found in exosome-enriched samples, but this can vary depending on source, isolation method, and/or EV biogenesis mechanism. The MILLIPLEX® Human Exosome Characterization panel provides a novel tool for researchers to study exosomal makers and content in lysed exosome samples.

Research category: Cancer

Research sub-category:
Inflammation & Autoimmune Mechanisms
Inflammation
Infectious Diseases
Immune Response
Tumor Markers

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel, except Syntenin-1 and Flotillin-1 antibodies had minimal cross reactivity with GAPDH standard.

Application

MILLIPLEX® Human Exosome Characterization Magnetic Bead Panel is a 9-plex kit to be used for the simultaneous quantification of any or all of the following analytes in exosomes isolated from serum, plasma and tissue culture supernatants: Argonaute-2 (Ago2), Calreticulin (CALR), CD9, CD63, CD81, Flotillin-1 (FLOT-1), GAPDH, Syntenin-1 (Syn-1), and TSG101.

  • This is an overnight assay.
  • Each assay requires 25 µL per well of diluted or neat isolated exosomes.
  • Argonaute-2 (Ago2) is a qualitative assay with the MCF7:IGF-1 Cell Lysate included as a control. All other analytes are for quantitative analysis.
  • To test the purity of EV preparation, the MILLIPLEX® Human Apolipoprotein 6-Plex Panel (Cat. No. APOMAG-62K) can be used.
  • To asses the functional component of EVs, the MILLIPLEX® Human Cytokine/Chemokine Growth Factor Panel A (Cat. No. HCYTA-60K), Panel B (HCYTB-60K), and Multi-Species TGFβ Panel (Cat. No. TGFBMAG-64K-03) can be used.
  • Following sample collection and preparation, exosomes can now be isolated and lysed according to your lab’s established protocol. We recommend Cell Signaling Lysis Buffer (Cat. No. 43-040).
  • Each lab may use its preferred lysis buffer but must predetermine that the assay will not be negatively affected by the lysis buffer.

Features and Benefits

This is a configurable kits. Design your multiplex kit by choosing available analytes within this panel. This panel is available for custom bead premixing.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service