Skip to Content
Merck
All Photos(1)

Key Documents

GR01L

Sigma-Aldrich

Anti-EGFR (Ab-1) Mouse mAb (528)

lyophilized, clone 528, Calbiochem®

Synonym(s):

Anti-Epidermal Growth Factor Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

528, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

should not react with

rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2a

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)

General description

Anti-EGFR (Ab-1), mouse monoclonal, clone 528, recognizes EGF receptor protein in A431 cells and normal skin. It is validated for use in ELISA, IF, IP, IHC (frozen,paraffin) & neutralization studies.
Purified mouse monoclonal antibody generated by immunizing Balb/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells (see application references). Recognizes the ~170 kDa EGFR protein.
Recognizes the EGF receptor protein in A431 cells and normal skin.

Immunogen

partially purified, human EGFR from human A431 cells

Application

Flow Cytometry (5 µg/ml)

Frozen Sections (2.5 µg/ml)

Immunofluorescence (5 µg/ml)

Immunoprecipitation (1 µg/sample)

Neutralization Studies (see comments and application references)

Paraffin Sections (see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM ammonium bicarbonate solution, 0.1 mg FAF-BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile phosphate buffered saline (PBS), pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml; product will be more stable if 0.1% sodium azide is included (do not add azide if antibody is to be used with viable cells). Lyophilized antibody should be resuspended at 4°C with occasional gentle mixing for at least 2 h. Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage.

Analysis Note

Positive Control
A-431 cells or skin tissue

Other Notes

Hunter T. 1984. Nature311, 414.
Masui, H., et al. 1984. Cancer Res.44, 1002.
Reynolds, F. H., Jr., et al. 1981. Nature 292, 259.
Ushiro, H. and Cohen, S. 1980. J. Biol. Chem.255, 8363.
This antibody binds to A431, HeLa, and other human cells but not to rodent cells. This antibody inhibits EGF binding to its receptor and is an antagonist of in vivo EGF-stimulated tyrosine kinase activity. Also inhibits A431 tumor formation in nude mice. For neutralization studies, use Cat. No. GR01L; Cat. No. GR01 contains azide, which is toxic to cells. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aidan P McCann et al.
Cell communication and signaling : CCS, 16(1), 77-77 (2018-11-10)
The deubiquitinase USP17 is overexpressed in NSCLC and has been shown to be required for the growth and motility of EGFR wild-type (WT) NSCLC cells. USP17 is also required for clathrin-mediated endocytosis of EGFR. Here, we examine the impact of
R Bauske et al.
Digestion, 61(4), 230-236 (2000-07-06)
Polyamines (putrescine, spermidine and spermine) are ubiquitous molecules indispensable for cell proliferation. In the intestinal lumen they are present in high amounts. Polyamine accumulation in proliferating cells of the intestinal mucosa is high, and it occurs both by enhanced synthesis
Joanne Morrison et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 16(2), 244-251 (2007-12-12)
Adenovirus gene therapy for intraperitoneal (IP) cancer is limited in clinical trials by inefficient tumor cell transduction and development of peritoneal adhesions. We have shown previously that normal virus tropism can be ablated by physically shielding the virus surface with
Alexia Conte et al.
Methods in molecular biology (Clifton, N.J.), 1652, 81-100 (2017-08-10)
Ubiquitination of the epidermal growth factor receptor (EGFR) is an important intracellular signal that occurs upon EGF stimulation and controls EGFR trafficking at multiple steps, finally destining the receptor to lysosomal degradation. In this chapter, we give an overview of
Jakub Jaworski et al.
Oncotarget, 5(16), 6964-6975 (2014-07-16)
Previously we have shown that expression of the deubiquitinating enzyme USP17 is required for cell proliferation and motility. More recently we reported that USP17 deubiquitinates RCE1 isoform 2 and thus regulates the processing of 'CaaX' motif proteins. Here we now

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service