GF327
IL-15,
Human Recombinant Animal Free
Synonym(s):
Interleukin-15, IL15
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77
Recommended Products
General description
IL-15 is an immunomodulating cytokine that stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. IL-15 exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. Increased expression of IL-15 has been implicated with rheumatoid arthritis, inflammatory bowl disease and diseases affiliated with retroviruses HIV and HTLV-I.
Recombinant human IL-15 is a 12.9 kDa protein consisting of 115 amino acid residues.
Recombinant human IL-15 is a 12.9 kDa protein consisting of 115 amino acid residues.
Quality
The ED50 was determined by its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Physical form
Product is presented in 20 mM Sodium Phosphate, pH 8.0 and is filtered through a 0.2 micron filter before lyophilization.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service