FR180380625
Estapor® Fluorescent Carboxylated microspheres
F1-EU-050 FluorEU COOH PS 1%, Red Fluorescent carboxylated microspheres 500nm, Lateral Flow Immunoassay - For ordering, click "Request more information"
Synonym(s):
Detector Particles, Fluorescent Beads
About This Item
Recommended Products
Quality Level
packaging
bottle of 10-1000 mL
manufacturer/tradename
Estapor®
concentration
1% solids
technique(s)
lateral flow assay: suitable (Immunoassay)
color
fluorescent red
matrix
polymeric Poly-styrene-co-acrylate
bead diameter
0.450-0.530 μm±<5%
capacity
1-10 μeq/g surface charge density
λmax
350 nm (Emission at 615 nm)
functional group
carboxylic acid (COOH)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service