Skip to Content
Merck
All Photos(1)

Key Documents

FCABS400F

Sigma-Aldrich

Milli-Mark® Anti-FcεRI Antibody, γ subunit-FITC

Milli-Mark®, from rabbit

Synonym(s):

Fc fragment of IgE high affinity I receptor for gamma polypeptide, Fc-epsilon RI-gamma, FceRI gamma, IgE Fc receptor subunit gamma, Immunoglobulin E receptor, high affinity,gamma chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FCER1G(2207)

General description

Type I allergic reactions are mediated through the IgE high affinity receptor (FceRI). When a multivalent antigen is presented, a redistribution of the monomeric IgE FceRI receptor complexes cause the degranulation of mast cells and basophils, resulting in the subsequent release of factors responsible for type I allergic reactions. The FceRI receptor is a tetrameric complex, consisting of an a-chain, b-chain and a dimeric g-chain. While the a-chain is primarily involved with the binding of IgE, the signal is transferred through the g subunit, which contains the immunoreceptor tyrosine activation motifs (ITAM) critical for initiating receptor mediated signal transduction through the Syk and Lyn tyrosine kinases. The FceRI receptor is able to engage and disengage the IgE, providing a versatile model for studying the function of kinase coupled receptors.

Specificity

Recognizes and is specific for FcεRI, γ-subunit.

Immunogen

Synthetic peptide corresponding to amino acids 80-86 of γ-subunit of Fc receptors.

Application

Milli-Mark Anti-FcεRI Antibody, γ subunit-FITC is an antibody against FcεRI for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

Evaluated by flow cytometry using RBL cells.

Target description

10 kDa Calculated

Physical form

Protein A purified
Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
RBL cells

Other Notes

Optimal working dilutions must be determined by end user for other instruments
Unconjugated antibody (Cat No. 06-727) has been shown to work on Human and Mouse.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spandan V Shah et al.
Cell reports, 25(10), 2766-2774 (2018-12-06)
Despite burgeoning evidence demonstrating the adaptive properties of natural killer (NK) cells, mechanistic data explaining these phenomena are lacking. Following antibody sensitization, NK cells lacking the Fc receptor (FcR) signaling chain (Δg) acquire adaptive features, including robust proliferation, multifunctionality, rapid
Marc Potempa et al.
Journal of immunology (Baltimore, Md. : 1950), 208(7), 1742-1754 (2022-03-25)
Although interactions between inhibitory Ly49 receptors and their self-MHC class I ligands in C57BL/6 mice are known to limit NK cell proliferation during mouse CMV (MCMV) infection, we created a 36-marker mass cytometry (CyTOF) panel to investigate how these inhibitory
Zeguang Wu et al.
Journal of immunology (Baltimore, Md. : 1950), 207(10), 2534-2544 (2021-10-10)
Human CMV (HCMV) is a ubiquitous pathogen that indelibly shapes the NK cell repertoire. Using transcriptomic, epigenomic, and proteomic approaches to evaluate peripheral blood NK cells from healthy human volunteers, we find that prior HCMV infection promotes NK cells with
James D Dahlvang et al.
Journal of immunology (Baltimore, Md. : 1950), 210(8), 1108-1122 (2023-03-08)
CMV infection alters NK cell phenotype and function toward a more memory-like immune state. These cells, termed adaptive NK cells, typically express CD57 and NKG2C but lack expression of the FcRγ-chain (gene: FCER1G, FcRγ), PLZF, and SYK. Functionally, adaptive NK
Cristina Capuano et al.
Frontiers in immunology, 9, 1031-1031 (2018-06-06)
Natural killer (NK) cells represent a pivotal player of innate anti-tumor immune responses. The impact of environmental factors in shaping the representativity of different NK cell subsets is increasingly appreciated. Human cytomegalovirus (HCMV) infection profoundly affects NK cell compartment, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service