Skip to Content
Merck
All Photos(1)

Key Documents

EZML

Millipore

Mouse Leptin ELISA Kit

measures and quantifies Leptin levels in 10 μL serum or plasma

Synonym(s):

Leptin, Obesity factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Product Name

Mouse Leptin ELISA, This Mouse Leptin ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine & Neuroscience research.

Quality Level

species reactivity

mouse

packaging

kit of 1 × 96 wells

parameter

10 μL sample volume (4hr assay)

assay range

accuracy: 87-100%
linearity: 100-111%
sensitivity: 0.05 ng/mL
standard curve range: 0.2-30 ng/mL

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma (K2 EDTA)

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Mouse Leptin ELISA is used to measure and quantify leptin levels in metabolism, endocrine, and neuroscience research. Leptin is a hormone or cytokine derived from adipocytes circulating in the blood. It acts as a regulator of food intake and energy expenditure via the hypothalamus. This hormone plays a role in angiogenesis, lipid and carbohydrate metabolism, and hematopoiesis. Leptin exerts its effects on the cardiovascular, reproductive, and immune systems. The deficiency of leptin in rodents is associated with obesity and insulin resistance.

Application

Mouse Leptin ELISA has been used to determine the levels of leptin in mice serum and plasma.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of chronic and intermittent calorie restriction on serum adiponectin and leptin and mammary tumorigenesis.
Rogozina OP, Bonorden MJ, Seppanen CN, Grande JP, Cleary MP
Cancer Prevention Research (Philadelphia, Pa.) null
Effects of castration on insulin levels and glucose tolerance in the mouse differ from those in man.
Takamitsu Inoue,Mahvash Zakikhani,Stephanie David,Carolyn Algire,Marie-Jose Blouin,Michael Pollak
Prostate null
Leptin signalling
Sweeney G
Cellular Signalling, 14(8), 655-663 (2002)
Exogenous insulin enhances humoural immune responses in short-day, but not long-day, Siberian hamsters (Phodopus sungorus).
Garcia NW, Greives TJ, Zysling DA, French SS, Chester EM, Demas GE
Proceedings. Biological Sciences / The Royal Society null
Candela Caballero-Eraso et al.
The Journal of physiology, 597(1), 151-172 (2018-10-05)
Leptin is a potent respiratory stimulant. A long functional isoform of leptin receptor, LepRb , was detected in the carotid body (CB), a key peripheral hypoxia sensor. However, the effect of leptin on minute ventilation (VE ) and the hypoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service