Skip to Content
Merck
All Photos(1)

Documents

EZHPYYT

Millipore

Human PYY ELISA Kit

measures and quantifies total PYY levels in 20 μL serum or plasma

Synonym(s):

PYY ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

product name

Human PYY (Total) ELISA, This Human PYY (Total) ELISA is used to measure & quantify PYY (Total) levels in Metabolism & Endocrine & Neuroscience research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

assay range

accuracy: 83.3-88.9%
(Serum)

accuracy: 95.9-102.5%
(Plasma)

linearity: 94-99%
(plasma)

linearity: 97-105%
(serum)

sensitivity: 1.4 pg/mL
standard curve range: 10-2,000 pg/mL
inter-assay cv: 3.7-16.5%
intra-assay cv: 0.9-5.8%
(Plasma)

inter-assay cv: 6.1-6.9%
intra-assay cv: 1.5-2.7%
(Serum)

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Peptide YY (PYY) is a hormone released by the lower GI tract with circulating levels increasing after meals in direct proportion to the caloric content. It is one of the major physiological signals to reduce appetite and food intake. Other functions of PYY are the inhibition of gastric acid secretion, GI motility and pancreatic enzyme output.

Specificity

Human PYY 3-36 100%~Human PYY 1-36 104%~Human [Leu31,Pro34] PYY 138%~Human [Pro34] PYY 158%~Porcine, Canine, Rat, or Mouse PYY Not Detectable

Application

3 hour assay using 20 µL sample size ~ Room Temperature
Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Used to detect/quantify: PYY (Total)

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptide tyrosine tyrosine levels are increased in patients with urea cycle disorders.
Sabrina Mitchell,Teresa Welch-Burke,Logan Dumitrescu,Jefferson P Lomenick et al.
Molecular Genetics and Metabolism null
Increased meal frequency does not promote greater weight loss in subjects who were prescribed an 8-week equi-energetic energy-restricted diet.
Jameason D Cameron,Marie-Josee Cyr,Eric Doucet
The British Journal of Nutrition null
Peptide YY (PYY) gene polymorphisms in the 3'-untranslated and proximal promoter regions regulate cellular gene expression and PYY secretion and metabolic syndrome traits in vivo.
Pei-An Betty Shih,Lei Wang,Stephane Chiron,Gen Wen,Caroline Nievergelt,Manjula Mahata et al.
The Journal of clinical endocrinology and metabolism null
Increases in peptide Y-Y levels following oat beta-glucan ingestion are dose-dependent in overweight adults.
EJ Beck, LC Tapsell, MJ Batterham, SM Tosh, XF Huang
Nutrition Research (New York, N.Y.) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service