EZHAWG474
EZ-Pak® Membrane Filter
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, gridded white filter, suitable for bioburden testing, input: beverage(s), bottled water
Synonym(s):
Membrane filters, Semi-permeable membranes
About This Item
Recommended Products
material
gridded filter
gridded white filter
mixed cellulose esters (MCE)
Quality Level
Agency
ISO 7704
description
General microbiological analysis
sterility
sterile; ethylene oxide treated
feature
easily wetted with water
hydrophilic
individually wrapped
manufacturer/tradename
EZ-Pak®
filter diam.
47 mm
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type beverage(s)
sample type bottled water
sample type soft drink
sample type sport drink
sample type cosmetics
sample(s) (environmental)
sample type pharmaceutical(s)
sample type beer
sample type wine
sample type food(s)
sample type cider
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical
compatibility
for use with EZ-Fit® Filtration Unit
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service