Skip to Content
Merck
All Photos(2)

Documents

EZFITSAM6

Millipore

EZ-Fit® Manifold

6-place manifold For EZ-Fit® filtration units, stainless steel

Synonym(s):

EZ-Fit® Filtration Manifold, EZ-Fit® manifold for EZ-Stream® vacuum pump, high throughput application, Filtration Manifold Base, Manifold Base, Manifold head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

316L stainless steel

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

pharmaceutical

compatibility

for use with EZ-Fit® Manifold Filtration system.

Looking for similar products? Visit Product Comparison Guide

General description

EZ-Fit® Filtration Manifold Base with 6 places is uniquely designed to carry out laboratory filtration of samples. The filtration manifold construction eliminates all sources of contamination over repeated use as each part of the manifold base is cleanable and autoclavable.
The EZ-Fit® Filtration Manifold Base can accommodate six filtration heads at a time, which would mean six samples can be tested at one go, and the ergonomic design enables operation under laminar flow hood and save laboratory bench space.
The 2-way valve system of the filtration manifold prevents the user to accidentally touch the filtration head while opening or closing the valve.

Application

EZ-Fit® Filtration Manifold Base with 6 places is used to carry out routine laboratory filtration samples. Liquid samples of raw materials, in process and finished products from diverse industries can be tested using the equipment.

Features and Benefits

The following features and benefits are associated with EZ-Fit® Filtration Manifold Base:
  • Different filtration heads fit both reusable and disposable filtration devices.
  • Each component is removable for deeper cleaning.
  • Autoclavable parts.
  • Quick fit connections for vacuum tubing.
  • Low height for easier use inside Laminar Flow Hood.

Packaging

EZFITBASE6 x1
EZFITMHTA3 x2

Analysis Note

The EZ-Fit® Filtration Manifold Base with 6 places can filter 6 test samples at once.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

configured for

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service