Skip to Content
Merck
All Photos(1)

Documents

ELI-008-M

Sigma-Aldrich

IL-6 ELISPOT Antibody Pair, Mouse

from rat, clone MP5-20F3, clone MP5-32C11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

clone

MP5-20F3, monoclonal
MP5-32C11, monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable

shipped in

wet ice

General description

Interleukin 6 (IL-6) is a multifunctional cytokine that plays an important role in regulating immune and inflammatory responses. Produced by a wide variety of cells, including T cells, monocytes, fibroblasts, endothelial cells, and keratinocytes, IL-6 works by stimulating B-cell differentiation and antibody production, as well inducing expression of hepatic acute-phase proteins, regulating bone metabolism, and promoting neuronal differentiation. In addition, IL-6 appears to interact with IL-2 in the proliferation of T-lymphocytes and with IL-3 to promote the proliferation of multipotent hematopoietic progenitor cells. The Millipore mouse IL-6 ELISPOT Antibody Pair provides a sensitive and highly reliable method for the detection of IL-6 secreting cells at the single cell level.

Specificity

Both the capture and the detection antibodies included in this kit react with mouse interleukin -6 (IL-6).

Application

IL-6 ELISPOT Antibody Pair, Mouse is a highly specific rat monoclonal antibody & that targets IL-6 & has been tested in ELISA.
Research Category
Inflammation & Immunology

Packaging

Sufficient reagents for five, 96-well plates.

Components

• Mouse IL-6 Capture Antibody, clone MP5-20F3. Quantity: 500 µg.

• Mouse IL-6 Detection Antibody, clone MP5-32C11. Quantity: 100 µg.

• Streptavidin Alkaline Phosphatase. Quantity: 55 μL.

Target description

~25 kDa

Physical form

Format: Purified
The 500 µg purified IL-6 capture antibody is buffered in a preservative-free phosphate solution, pH 7.2, and the 100 µg purified IL-6 biotinylated detection antibody is buffered in a phosphate solution, pH 7.2, containing 0.09% sodium azide. Streptavidin alkaline phosphatase is supplied in a Tris Buffer, pH 8.0, containing 0.1 M sodium chloride, 1 mM magnesium chloride, 0.2% (w/v) BSA, 10% (w/w) glycerol, and 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for up to six months from the date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service