ABN240
Anti-Beta (β)-Amyloid antibody
rabbit polyclonal
Synonym(s):
Amyloid beta A4 protein, ABPP, APPI, APP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II, 1.N-APP2.Soluble APP-alpha, S-APP-alpha3.Soluble APP-beta, S-APP-beta4.C995.Beta-amyloid protein 42, Be
About This Item
Recommended Products
Product Name
Anti-Amyloid, β 1-40, aβ Antibody, from rabbit, purified by affinity chromatography
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
bovine (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology)
technique(s)
ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... APP(351)
General description
Specificity
Immunogen
Application
ELISA: A representative lot detected Amyloid, B 1-40 in ELISA (Direct).
Neuroscience
Neurodegenerative Diseases
Quality
Western Blot Analysis: 0.5 µg/mL of this antibody detected Amyloid, B 1-40 on 10 µg of human Alzheimer brain tissue lysate.
Target description
Linkage
Physical form
Storage and Stability
Analysis Note
Human Alzhiemer brain tissue lysate
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service