Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
2
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, mouse
species reactivity (predicted by homology)
nonhuman primates (based on 100% sequence homology), feline (based on 100% sequence homology), canine (based on 100% sequence homology), rat (based on 100% sequence homology), hamster (based on 100% sequence homology)
technique(s)
immunocytochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CASP3(836)
General description
Caspase-3 (EC 3.4.22.56; UniProt P42574; also known as Apopain, CASP-3, CPP-32, PARP cleavage protease, SCA-1, SREBP cleavage activity 1, Caspase 3 apoptosis-related cysteine protease, Cysteine protease CPP32, Procaspase3, Protein Yama) is encoded by the CASP3 (also known as CPP32, CPP32B) gene (Gene ID 836) in human. Cysteine-aspartic proteases or Caspases play essential roles in apoptosis, necrosis, and inflammation. Historically, caspases were numbered in the order in which they were identified. Caspase-3 is initially produced with a propeptide sequence (a.a. 1-9), the removal of which yields the 268 a.a. caspase-3 proenzyme. Upon activation, the proenzyme is proteolytically cleaved first between Asp175-Ser176 to generate a p20 (a.a. 10-175) fragment and the p12 (a.a. 176-277) subunit. Further cleavage of the p20 fragment between Asp28-Ser29 produces the p17 (a.a. 29-175) subunit. The p17 and p12 dimerizes and forms the active caspase-3 enzyme. Caspase-3 plays a central role in the sequential activation of caspases during the execution phase of cell apoptosis. Caspase-3 mediates the proteolytic activation of caspases-6 and -7, while caspase-3 itself is processed and activated by caspase-8, -9, or -10. Caspase-3 is the predominant caspase involved in the cleavage of amyloid-beta 4A precursor protein, which is associated with neuronal death in Alzheimer′s disease.
~32 kDa observed
Immunogen
Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human Caspase 3 near the C-terminus.
Application
Anti-Caspase 3 Antibody is an antibody against Caspase 3 Antibody for use in Western Blotting and Immunocytochemistry.
Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected Caspase 3 in HeLa, A431, HUVEC, and NIH/3T3 cells.
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
Biochem/physiol Actions
Expected to react with pro-caspase 3, the uncleaved/inactive caspase 3 (32 kDa), and the p12 (12 kDa), but not the p17 (17 kDa) subunit of cleaved/active caspase 3.
Physical form
Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in A431 cell lysate.
Western Blotting Analysis: 0.05 µg/mL of this antibody detected Caspase 3 in 10 µg of A431 cell lysate.
Western Blotting Analysis: 0.05 µg/mL of this antibody detected Caspase 3 in 10 µg of A431 cell lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Replaces: 06-375
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Caspase 3 Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.