Skip to Content
Merck
All Photos(1)

Key Documents

AB5977

Sigma-Aldrich

Anti-Musashi-1 Antibody

Chemicon®, from rabbit

Synonym(s):

Musashi homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rodent, mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MSI1(4440)

General description

In mammals, the Musashi family is important for cell fate determination, playing roles in maintenance of the stem-cell state, differentiation and tumorigenesis. Musashi1 (also known as Msi1), is selectivley expressed in neural progenitor cells, including neural stem cells. Outside the nervous system, Musashi1 is a selective marker for intestinal stem or early lineage cells. Musashi1 interacts with the Notch pathway during asymmetric cell division by binding the 3′ UTR of Numb. Numb is prevented from repressing Notch signaling when Musashi1 is present.

Specificity

Reactivity with other species has not been confirmed.
Recognizes Musashi-1.

Immunogen

Synthetic peptide, amino acids 5-21 of Musashi.

Application

Detect Musashi-1 using this Anti-Musashi-1 Antibody validated for use in IC, IH & WB.
Immunohistochemistry:
1:200-1:1,000 dilution from a previous lot was used.

Immunocytochemistry:
1:200-1:1,000 dilution from a previous lot was used.

Western blot:
1:200-1:1,000 using ECL.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Developmental Neuroscience

RNA Binding Protein (RBP)

Quality

Routinely evaluated by Western Blot on Human Placenta lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected Musashi-1 on 10 μg of Human placenta lysates.

Target description

39 kDa

Linkage

Replaces: 03-114; 04-1041

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal in buffer containing a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at -80ºC from date of receipt.

Analysis Note

Control
Mouse cortical neural stem cells (Catalog No. SCR029).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Musashi1-CreER(T2) : a new cre line for conditional mutagenesis in neural stem cells.
Takeda, H; Koso, H; Tessarollo, L; Copeland, NG; Jenkins, NA
Genesis (2000)
Raffaella Scardigli et al.
Stem cells (Dayton, Ohio), 32(9), 2516-2528 (2014-05-09)
Adult neurogenesis is a multistep process regulated by several extrinsic factors, including neurotrophins. Among them, little is known about the role of nerve growth factor (NGF) in the neurogenic niches of the mouse. Here we analyzed the biology of adult
Cell lineage identification and stem cell culture in a porcine model for the study of intestinal epithelial regeneration.
Gonzalez, LM; Williamson, I; Piedrahita, JA; Blikslager, AT; Magness, ST
Testing null
Mara Vinci et al.
Nature medicine, 24(8), 1204-1215 (2018-07-04)
The failure to develop effective therapies for pediatric glioblastoma (pGBM) and diffuse intrinsic pontine glioma (DIPG) is in part due to their intrinsic heterogeneity. We aimed to quantitatively assess the extent to which this was present in these tumors through
Alexi Nott et al.
Nature neuroscience, 19(11), 1497-1505 (2016-10-28)
Mutations in MECP2 cause the neurodevelopmental disorder Rett syndrome (RTT). The RTT missense MECP2R306C mutation prevents MeCP2 from interacting with the NCoR/histone deacetylase 3 (HDAC3) complex; however, the neuronal function of HDAC3 is incompletely understood. We found that neuronal deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service