Skip to Content
Merck
All Photos(1)

Key Documents

AB3542

Sigma-Aldrich

Anti-Monocarboxylate Transporter 2 Antibody

serum, Chemicon®

Synonym(s):

MCT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A7(9194)

Specificity

Recognizes rat Monocarboxylate Transporter 2 (MCT2). The immunogen shows no significant sequence homology with other MCT.

The immunogen sequence is 93% conserved in mouse and 66% in hamster. Reactivity with other species has not been confirmed.

Immunogen

A 15 amino acid peptide sequence near the C-terminus of rat MCT2 (Jackson et al. 1997; Dao et al. 1998; Koehler-Stec et al. 1998; Price et al. 1998; Halestrap & Price 1999).

Application

Anti-Monocarboxylate Transporter 2 Antibody is an antibody against Monocarboxylate Transporter 2 for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog number AG218) per well.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3542P

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogen Receptor Involvement in Noradrenergic Regulation of Ventromedial Hypothalamic Nucleus Glucoregulatory Neurotransmitter and Stimulus-Specific Glycogen Phosphorylase Enzyme Isoform Expression.
A S M H Mahmood et al.
ASN neuro, 12, 1759091420910933-1759091420910933 (2020-04-03)
J S Gilbert et al.
Placenta, 33(2), 100-105 (2011-12-22)
While utero-placental insufficiency is associated with adverse outcomes for both mother and fetus, many of the maternal-fetal adaptations during pregnancy in models of fetal compromise remain unclear. The purpose of this study was to determine if chronically reduced uterine perfusion
Estrogen Receptor Involvement in Noradrenergic Regulation of Ventromedial Hypothalamic Nucleus Glucoregulatory Neurotransmitter and Stimulus-Specific Glycogen Phosphorylase Enzyme Isoform Expression.
A S M H Mahmood et al.
ASN neuro, 12, 1759091420910933-1759091420910933 (2020-04-03)
E M England et al.
Journal of animal science, 95(8), 3552-3562 (2017-08-15)
Acute activation of AMP-activated protein kinase (AMPK) increases monocarboxylate transporter (MCT) expression in skeletal muscle. However, the impact of chronic activation of AMPK on MCT expression in skeletal muscle is unknown. To investigate, MCT1, MCT2, and MCT4 mRNA expression and
Emmanuel Cruz et al.
Glia, 70(11), 2207-2231 (2022-08-03)
The consumption of glucose in the brain peaks during late childhood; yet, whether and how glucose metabolism is differentially regulated in the brain during childhood compared to adulthood remains to be understood. In particular, it remains to be determined how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service