Skip to Content
Merck
All Photos(1)

Key Documents

AB3402P

Sigma-Aldrich

Anti-Agouti Related Protein Antibody

Chemicon®, from rabbit

Synonym(s):

AGRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
577428-U577420-U577421-U
matrix active group

bonding phase, Diphenyl (diphenylmethyl)

matrix active group

bonding phase, Diphenyl (diphenylmethyl)

matrix active group

bonding phase, Diphenyl (diphenylmethyl)

matrix active group

bonding phase, Diphenyl (diphenylmethyl)

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

particle size

2.7 μm

particle size

2.7 μm

particle size

2.7 μm

particle size

2.7 μm

pore size

1000 Å pore size

pore size

1000 Å pore size

pore size

1000 Å pore size

pore size

1000 Å pore size

product line

BIOshell

product line

BIOshell

product line

BIOshell

product line

BIOshell

General description

Agouti-related peptide-expressing (AgRP) neurons seem to be critical for the acute regulation of eating. The AgRP protein is a strong antagonis of MC3-R and MC4-R and is expressed primarily in the adrenal gland, subthalamic nucleus, and hypothalamus, with a lower level of expression occurring in testis, lung, and kidney.

Specificity

Recognizes AGRP. The immunogen shows no significant sequence homology with Agouti, tubby, tub, TULP1 or TULP2.

SPECIES REACTIVITIES: The immunogen sequence is 85% homologous with human and bovine. Reactivity with other species has not been confirmed.

Immunogen

A 15 amino acid peptide near the C-terminus of mouse AGRP (Shutter et al. 1997; Ollmann et al. 1997).

Application

Detect Agouti Related Protein using this Anti-Agouti Related Protein Antibody validated for use in ELISA & WB.
Research Category
Metabolism
Research Sub Category
Glucose/Glycogen Metabolism
Western blot: 1-10 μg/mL using ECL.



ELISA: 1:10,000-1:50,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in phosphate buffered saline containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service