Skip to Content
Merck
All Photos(1)

Key Documents

AB1924

Sigma-Aldrich

Anti-Integrin α4 Antibody, cytoplasmic on CT

serum, Chemicon®

Synonym(s):

CD49d

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, chicken, hamster, rat, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

chicken ... Itga4(424121)
human ... ITGA4(3676)
mouse ... Itga4(16401)
pig ... Itga4(100521477)
rat ... Itga4(311144)

Specificity

Integrin alpha 4. Specificity determined by immunoprecipitation from a detergent extract of 35S methionine-labeled lymphoid cells. No cross-reactivity to alpha 1, alpha 2, alpha 3, alpha 5, alpha 6 or alpha V. The antibody recognizes an epitope on the cytoplasmic domain of the protein, and it is thus intracellular.

Immunogen

Epitope: cytoplasmic domain, C-terminus
Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human alpha 4 integrin subunit.

Application

Anti-Integrin α4 Antibody, cytoplasmic on C-terminus is an antibody against Integrin α4 for use in ELISA, IP, RIA, WB, IH.
ELISA/RIA: 1:1000.

Western blot: 1:1000, non-reduced conditions; blocking should be with 1-3% BSA. Recognizes a 140 kDa human alpha 4 band under non-reducing conditions.

Immunohistochemistry: 1:500

Immunoprecipitation: 5 μL of antibody is sufficient to precipitate alpha4 from 5x106 cells.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Linkage

Replaces: 04-1131

Physical form

Liquid rabbit antiserum with 0.05% sodium azide.

Storage and Stability

Maintain at -20ºC in undiluted aliquots for up to 12 months upon date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Sens et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 32(1), 70-81 (2016-07-19)
Patients with cholestatic liver disease experience increased fracture risk. Higher circulating levels of a fibronectin isoform called oncofetal fibronectin (oFN) were detected in a subset of such patients. Administering this isoform to mice suppresses osteoblast differentiation and diminishes bone mineral
Carla Sens et al.
The Journal of biological chemistry, 292(19), 7745-7760 (2017-03-23)
Fibronectin is a multidomain protein secreted by various cell types. It forms a network of fibers within the extracellular matrix and impacts intracellular processes by binding to various molecules, primarily integrin receptors on the cells. Both the presence of several
Fei Wang et al.
The Journal of clinical investigation, 120(11), 3979-3995 (2010-10-28)
Sphingosine-1-phosphate (S1P) is a biologically active sphingolipid that has pleiotropic effects in a variety of cell types including ECs, SMCs, and macrophages, all of which are central to the development of atherosclerosis. It may therefore exert stimulatory and inhibitory effects
Miho Takahashi et al.
International journal of cancer, 121(9), 1919-1929 (2007-07-11)
It has been proposed that the metastasis suppressor CD82/KAI-1, which is a member of the tetraspanin superfamily, regulates biological activity by associating with cell surface receptors or proteins. We show a novel association between CD82 and the hepatocyte growth factor
Nasim Jamali et al.
FASEB bioAdvances, 1(7), 415-434 (2019-08-10)
We have previously demonstrated that the active form of vitamin D (calcitriol; 1,25(OH)2D3) is a potent inhibitor of retinal neovascularization. However, the underlying molecular and cellular mechanisms involved remained poorly understood. Perivascular supporting cells including pericytes (PC) play important roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service